Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01523227_m1
          See other ARHGAP29 GE Assays ›
          Gene Symbol:
          Arhgap29
          Gene Name:
          Rho GTPase activating protein 29
          Gene Aliases:
          B130017i01rik, RGD1306185
          Chromosome Location:
          Chr.2: 225552076 - 225613349 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01523227_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arhgap29
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009405.1 NP_001009405.1 23 - 24 3125 89
          XM_006233239.2 XP_006233301.1 22 - 23 4925 89
          XM_006233240.2 XP_006233302.1 22 - 23 3114 89
          XM_006233241.2 XP_006233303.1 22 - 23 3364 89
          GenBank mRNA BC087167.1 - 23 - 24 3125 7135494 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002110.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arhgap29

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular signal transduction
          positive regulation of GTPase activity
          GTPase activator activity
          PDZ domain binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P344730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726268 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157918 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139259 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105376 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23893 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49738 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P344730 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P344730 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P344730 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P344730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70070 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48535 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18583 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95325 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818725 - -
          Affymetrix Rat Gene 1.0 ST Array 10818716 - -
          Affymetrix Rat Gene 1.0 ST Array 10818737 - -
          Affymetrix Rat Gene 1.0 ST Array 10818729 - -
          Affymetrix Rat Gene 1.0 ST Array 10818726 - -
          Affymetrix Rat Gene 1.0 ST Array 10818727 - -
          Affymetrix Rat Gene 1.0 ST Array 10818718 - -
          Affymetrix Rat Gene 1.0 ST Array 10818734 - -
          Affymetrix Rat Gene 1.0 ST Array 10818728 - -
          Affymetrix Rat Gene 1.0 ST Array 10818722 - -
          Affymetrix Rat Gene 1.0 ST Array 10818730 - -
          Affymetrix Rat Gene 1.0 ST Array 10818720 - -
          Affymetrix Rat Gene 1.0 ST Array 10818731 - -
          Affymetrix Rat Gene 1.0 ST Array 10818735 - -
          Affymetrix Rat Gene 1.0 ST Array 10818721 - -
          Affymetrix Rat Gene 1.0 ST Array 10818741 - -
          Affymetrix Rat Gene 1.0 ST Array 10818738 - -
          Affymetrix Rat Gene 1.0 ST Array 10818739 - -
          Affymetrix Rat Gene 1.0 ST Array 10818724 - -
          Affymetrix Rat Gene 1.0 ST Array 10818736 - -
          Affymetrix Rat Gene 1.0 ST Array 10818740 - -
          Affymetrix Rat Gene 1.0 ST Array 10818732 - -
          Affymetrix Rat Gene 1.0 ST Array 10818717 - -
          Affymetrix Rat Gene 1.0 ST Array 10818723 - -
          Affymetrix Rat Gene 1.0 ST Array 10818733 - -
          Affymetrix Rat Gene 1.0 ST Array 10818719 - -
          Affymetrix Rat Gene 1.1 ST Array 10818715 - -
          Rat Genome 230 2.0 Array 1398397_at - -
          Rat Genome 230 2.0 Array 1398397_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200014534.rn.1 TC0200002110.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10818715 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22172932 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353948 - -
          RatRef-12 Expression BeadChip ILMN_69985 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline