Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01523784_m1
          See other ZFP667 GE Assays ›
          Gene Symbol:
          Zfp667
          Gene Name:
          zinc finger protein 667
          Gene Aliases:
          Mip1, Znf667
          Chromosome Location:
          Chr.1: 71270101 - 71290365 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01523784_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp667
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008557.1 NP_001008557.1 4 - 5 363 116
          XM_006228189.2 XP_006228251.1 4 - 5 702 116
          GenBank mRNA AY221750.1 - 4 - 5 363 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004760.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp667

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          nucleic acid binding
          DNA binding
          transcription cofactor activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P746666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52277 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P746666 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P746666 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P746666 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P746666 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P130684 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6420 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54606 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718697 - -
          Affymetrix Rat Gene 1.0 ST Array 10718701 - -
          Affymetrix Rat Gene 1.0 ST Array 10718702 - -
          Affymetrix Rat Gene 1.0 ST Array 10718699 - -
          Affymetrix Rat Gene 1.0 ST Array 10718698 - -
          Affymetrix Rat Gene 1.0 ST Array 10718700 - -
          Affymetrix Rat Gene 1.1 ST Array 10718696 - -
          Rat Genome 230 2.0 Array 1392676_at - -
          Rat Genome 230 2.0 Array 1376482_at - -
          Rat Genome 230 2.0 Array 1376482_PM_at - -
          Rat Genome 230 2.0 Array 1392676_PM_at - -
          Affymetrix HT Rat Focus Array 1392676_PM_at - -
          Affymetrix HT Rat Focus Array 1376482_PM_at - -
          Affymetrix HT Rat Focus Array 1392676_at - -
          Affymetrix HT Rat Focus Array 1376482_at - -
          Affymetrix Rat Clariom S Assay PSR0100037412.rn.1 TC0100004760.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037416.rn.1 TC0100004760.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037414.rn.1 TC0100004760.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037418.rn.1 TC0100004760.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037419.rn.1 TC0100004760.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037417.rn.1 TC0100004760.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037415.rn.1 TC0100004760.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037413.rn.1 TC0100004760.rn.1 RTA TC
          Rat Expression Set 230 1376482_at - -
          Rat Expression Set 230 1376482_PM_at - -
          Rat Expression Set 230 1392676_PM_at - -
          Rat Expression Set 230 1392676_at - -
          Affymetrix Rat Gene 1.0 ST Array 10718696 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376265 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline