Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01524218_m1
          See other SFT2D1 GE Assays ›
          Gene Symbol:
          Sft2d1
          Gene Name:
          SFT2 domain containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 52998548 - 53014444 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01524218_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sft2d1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008302.1 NP_001008303.1 7 - 8 511 103
          GenBank mRNA BC085346.1 - 7 - 8 511 7304630 103
          FQ227930.1 - 7 - 8 489 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004503.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sft2d1

          Gene Ontology Categories:

          Function(s)

          protein transport
          vesicle-mediated transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P819377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131017 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P819377 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P819377 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P819377 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P819377 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7533 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90978 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15261 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718101 - -
          Affymetrix Rat Gene 1.0 ST Array 10718092 - -
          Affymetrix Rat Gene 1.0 ST Array 10718093 - -
          Affymetrix Rat Gene 1.0 ST Array 10718096 - -
          Affymetrix Rat Gene 1.0 ST Array 10718099 - -
          Affymetrix Rat Gene 1.0 ST Array 10718098 - -
          Affymetrix Rat Gene 1.0 ST Array 10718097 - -
          Affymetrix Rat Gene 1.0 ST Array 10718100 - -
          Affymetrix Rat Gene 1.0 ST Array 10718094 - -
          Affymetrix Rat Gene 1.1 ST Array 10718091 - -
          Rat Genome 230 2.0 Array 1373033_at - -
          Rat Genome 230 2.0 Array 1373033_PM_at - -
          Affymetrix HT Rat Focus Array 1373033_at - -
          Affymetrix HT Rat Focus Array 1373033_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100035698.rn.1 TC0100004503.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100035700.rn.1 TC0100004503.rn.1 RTA TC
          Rat Expression Set 230 1373033_PM_at - -
          Rat Expression Set 230 1373033_at - -
          Affymetrix Rat Gene 1.0 ST Array 10718091 - -
          Rat Genome U34 Set rc_AA900647_s_at - -
          Rat Genome U34 Set rc_AI235219_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21348549 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354859 - -
          RatRef-12 Expression BeadChip ILMN_63191 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline