Search
Search

Kirrel3| Gene Symbol: Kirrel3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048215.1 | NP_001041680.1 | 14 - 15 | 2076 | 59 | |
| XM_006242782.2 | XP_006242844.1 | 15 - 16 | 2206 | 59 | ||
| GenBank mRNA | DQ902828.1 | - | 14 - 15 | 2076 | 59 | |
Set Membership: |
|