Search
Search
| Gene Symbol: Ncr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181822.2 | NP_861543.2 | 2 - 3 | 570 | 77 | |
| XM_006256054.2 | XP_006256116.1 | 2 - 3 | 740 | 77 | ||
| GenBank mRNA | AY273824.1 | - | 2 - 3 | 691 | 77 | |
Set Membership: |
|