Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01526032_g1
          See other PRRC2A GE Assays ›
          Gene Symbol:
          Prrc2a
          Gene Name:
          proline-rich coiled-coil 2A
          Gene Aliases:
          Bat2
          Chromosome Location:
          Chr.20: 5139880 - 5155356 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01526032_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prrc2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_212462.2 NP_997627.1 29 - 30 6350 85
          XM_006256048.2 XP_006256110.1 31 - 32 6646 85
          XM_006256050.2 XP_006256112.1 31 - 32 6643 85
          XM_006256051.2 XP_006256113.1 30 - 31 6597 85
          XM_006256052.2 XP_006256114.1 30 - 31 6595 85
          XM_006256053.2 XP_006256115.1 30 - 31 6433 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001108.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prrc2a

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P065585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065585 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P065585 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20729 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828055 - -
          Affymetrix Rat Gene 1.0 ST Array 10828071 - -
          Affymetrix Rat Gene 1.0 ST Array 10828047 - -
          Affymetrix Rat Gene 1.0 ST Array 10828050 - -
          Affymetrix Rat Gene 1.0 ST Array 10828058 - -
          Affymetrix Rat Gene 1.0 ST Array 10828067 - -
          Affymetrix Rat Gene 1.0 ST Array 10828049 - -
          Affymetrix Rat Gene 1.0 ST Array 10828059 - -
          Affymetrix Rat Gene 1.0 ST Array 10828076 - -
          Affymetrix Rat Gene 1.0 ST Array 10828048 - -
          Affymetrix Rat Gene 1.0 ST Array 10828054 - -
          Affymetrix Rat Gene 1.0 ST Array 10828064 - -
          Affymetrix Rat Gene 1.0 ST Array 10828052 - -
          Affymetrix Rat Gene 1.0 ST Array 10828068 - -
          Affymetrix Rat Gene 1.0 ST Array 10828065 - -
          Affymetrix Rat Gene 1.0 ST Array 10828066 - -
          Affymetrix Rat Gene 1.0 ST Array 10828057 - -
          Affymetrix Rat Gene 1.0 ST Array 10828051 - -
          Affymetrix Rat Gene 1.0 ST Array 10828056 - -
          Affymetrix Rat Gene 1.0 ST Array 10828072 - -
          Affymetrix Rat Gene 1.0 ST Array 10828060 - -
          Affymetrix Rat Gene 1.0 ST Array 10828073 - -
          Affymetrix Rat Gene 1.0 ST Array 10828062 - -
          Affymetrix Rat Gene 1.0 ST Array 10828061 - -
          Affymetrix Rat Gene 1.0 ST Array 10828063 - -
          Affymetrix Rat Gene 1.0 ST Array 10828069 - -
          Affymetrix Rat Gene 1.0 ST Array 10828074 - -
          Affymetrix Rat Gene 1.0 ST Array 10828070 - -
          Affymetrix Rat Gene 1.0 ST Array 10828053 - -
          Affymetrix Rat Gene 1.0 ST Array 10828075 - -
          Affymetrix Rat Gene 1.1 ST Array 10828046 - -
          Rat Genome 230 2.0 Array 1388536_PM_at - -
          Rat Genome 230 2.0 Array 1388536_at - -
          Affymetrix HT Rat Focus Array 1388536_PM_at - -
          Affymetrix HT Rat Focus Array 1388536_at - -
          Affymetrix Rat Clariom S Assay PSR2000009276.rn.1 TC2000001108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000009278.rn.1 TC2000001108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000009277.rn.1 TC2000001108.rn.1 RTA TC
          Rat Expression Set 230 1388536_at - -
          Rat Expression Set 230 1388536_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828046 - -
          Rat Genome U34 Set rc_AI236583_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21470965 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357588 - -
          RatRef-12 Expression BeadChip ILMN_70076 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline