Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528166_g1
          See other PPP2R3B GE Assays ›
          Gene Symbol:
          Ppp2r3b
          Gene Name:
          protein phosphatase 2, regulatory subunit B'', beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 1469748 - 1476353 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528166_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp2r3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001139492.1 NP_001132964.1 12 - 13 2005 87
          GenBank mRNA BC161890.1 - 12 - 13 2005 7135877 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000025.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Protein Phosphatase 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          negative regulation of cell proliferation
          regulation of catalytic activity
          negative regulation of G1/S transition of mitotic cell cycle
          chromatin binding
          calcium ion binding
          protein phosphatase type 2A regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P22907 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22907 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P22907 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P22907 - -
          Array Name Probe IDs Targets Target Type
          Rat Genome 230 2.0 Array 1383081_PM_at - -
          Rat Genome 230 2.0 Array 1383081_at - -
          Rat Genome 230 2.0 Array 1396353_PM_at - -
          Rat Genome 230 2.0 Array 1396353_at - -
          Affymetrix HT Rat Focus Array 1383081_PM_at - -
          Affymetrix HT Rat Focus Array 1383081_at - -
          Affymetrix Rat Clariom S Assay PSR1400000163.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000171.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000162.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000167.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000169.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000160.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000161.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000170.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000164.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000168.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000165.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000166.rn.1 TC1400000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000159.rn.1 TC1400000025.rn.1 RTA TC
          Rat Expression Set 230 1383081_PM_at - -
          Rat Expression Set 230 1396353_at - -
          Rat Expression Set 230 1396353_PM_at - -
          Rat Expression Set 230 1383081_at - -
          Rat Genome U34 Set rc_AI177925_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21360588 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-2btvb:80/100.66.73.215:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline