Search
Search
| Gene Symbol: Cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199501.1 | NP_955795.1 | 1 - 2 | 261 | 63 | |
| XM_006240778.1 | XP_006240840.1 | 1 - 2 | 337 | 63 | ||
| GenBank mRNA | BC061832.1 | - | 1 - 2 | 261 | 5599608 | 63 |
| D28753.1 | - | 2 - 3 | 297 | 63 | ||
Set Membership: |
|