Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530527_m1
          See other FAM81A GE Assays ›
          Gene Symbol:
          Fam81a
          Gene Name:
          family with sequence similarity 81, member A
          Gene Aliases:
          RGD1311958
          Chromosome Location:
          Chr.8: 76521659 - 76579387 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01530527_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam81a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108163.1 NP_001101633.1 8 - 9 1331 76
          XM_006243378.2 XP_006243440.1 8 - 9 1167 76
          XM_006243379.2 XP_006243441.1 8 - 9 1239 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002863.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P885889 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P061817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118573 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87914 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146816 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134856 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13781 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P885889 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P885889 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061817 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061817 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P885889 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P502271 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P382523 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P885889 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60748 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918502 - -
          Affymetrix Rat Gene 1.0 ST Array 10918503 - -
          Affymetrix Rat Gene 1.0 ST Array 10918505 - -
          Affymetrix Rat Gene 1.0 ST Array 10918506 - -
          Affymetrix Rat Gene 1.0 ST Array 10918508 - -
          Affymetrix Rat Gene 1.0 ST Array 10918504 - -
          Affymetrix Rat Gene 1.0 ST Array 10918507 - -
          Affymetrix Rat Gene 1.1 ST Array 10918501 - -
          Rat Genome 230 2.0 Array 1394881_at - -
          Rat Genome 230 2.0 Array 1378245_PM_at - -
          Rat Genome 230 2.0 Array 1394881_PM_at - -
          Rat Genome 230 2.0 Array 1378245_at - -
          Affymetrix HT Rat Focus Array 1378245_PM_at - -
          Affymetrix HT Rat Focus Array 1394881_PM_at - -
          Affymetrix HT Rat Focus Array 1378245_at - -
          Affymetrix HT Rat Focus Array 1394881_at - -
          Affymetrix Rat Clariom S Assay PSR0800023124.rn.1 TC0800002863.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023123.rn.1 TC0800002863.rn.1 RTA TC
          Rat Expression Set 230 1394881_PM_at - -
          Rat Expression Set 230 1378245_at - -
          Rat Expression Set 230 1394881_at - -
          Rat Expression Set 230 1378245_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918501 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21951967 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367934 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530444 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline