Search
Search

RT1-T18| Gene Symbol: RT1-T18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002821.1 | NP_001002821.1 | 618 | 73 | ||
| XM_006255985.2 | XP_006256047.1 | 3 - 4 | 1185 | 73 | ||
| XM_006255986.2 | XP_006256048.1 | 3 - 4 | 1185 | 73 | ||
| XM_008772742.1 | XP_008770964.1 | 3 - 4 | 1185 | 73 | ||
| XM_008772743.1 | XP_008770965.1 | 4 - 5 | 650 | 73 | ||
| XM_008772744.1 | XP_008770966.1 | 4 - 5 | 1788 | 73 | ||
Set Membership: |
|