Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01533426_m1
          See other CD101 GE Assays ›
          Gene Symbol:
          Cd101
          Gene Name:
          CD101 molecule
          Gene Aliases:
          Igsf2
          Chromosome Location:
          Chr.2: 203376012 - 203413895 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01533426_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271217.1 NP_001258146.1 5 - 6 1669 70
          XM_006233049.2 XP_006233111.1 4 - 5 1254 70
          XM_008761362.1 XP_008759584.1 5 - 6 2439 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004326.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s)

          positive regulation of myeloid leukocyte differentiation

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P071725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071725 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P071725 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P489869 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10825447 - -
          Affymetrix Rat Gene 1.0 ST Array 10825450 - -
          Affymetrix Rat Gene 1.0 ST Array 10825448 - -
          Affymetrix Rat Gene 1.0 ST Array 10825451 - -
          Affymetrix Rat Gene 1.0 ST Array 10825445 - -
          Affymetrix Rat Gene 1.0 ST Array 10825449 - -
          Affymetrix Rat Gene 1.0 ST Array 10825446 - -
          Affymetrix Rat Gene 1.0 ST Array 10825444 - -
          Affymetrix Rat Clariom S Assay PSR0200029751.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029752.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029753.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029749.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029756.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029750.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029754.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029745.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029757.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029744.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029748.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029758.rn.1 TC0200004326.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10825443 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349278 - -
          RatRef-12 Expression BeadChip ILMN_58827 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline