Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01535579_m1
          See other TLE2 GE Assays ›
          Gene Symbol:
          Tle2
          Gene Name:
          transducin-like enhancer of split 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 10988536 - 11004860 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01535579_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tle2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039013.1 NP_001034102.1 13 - 14 1192 94
          XM_006240889.2 XP_006240951.1 13 - 14 1364 94
          XM_006240890.2 XP_006240952.1 13 - 14 1841 94
          XM_006240892.2 XP_006240954.1 13 - 14 1277 94
          XM_006240893.2 XP_006240955.1 13 - 14 1361 94
          XM_006240894.2 XP_006240956.1 13 - 14 1364 94
          XM_006240895.2 XP_006240957.1 13 - 14 1433 94
          GenBank mRNA BC100652.1 - 13 - 14 1192 7932424 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002099.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          negative regulation of transcription, DNA-templated
          negative regulation of canonical Wnt signaling pathway
          transcription corepressor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P140026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511721 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58371 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P140026 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P140026 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P140026 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P140026 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85642 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84403 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55993 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900180 - -
          Affymetrix Rat Gene 1.0 ST Array 10900177 - -
          Affymetrix Rat Gene 1.0 ST Array 10900173 - -
          Affymetrix Rat Gene 1.0 ST Array 10900178 - -
          Affymetrix Rat Gene 1.0 ST Array 10900183 - -
          Affymetrix Rat Gene 1.0 ST Array 10900184 - -
          Affymetrix Rat Gene 1.0 ST Array 10900185 - -
          Affymetrix Rat Gene 1.0 ST Array 10900172 - -
          Affymetrix Rat Gene 1.0 ST Array 10900174 - -
          Affymetrix Rat Gene 1.0 ST Array 10900176 - -
          Affymetrix Rat Gene 1.0 ST Array 10900165 - -
          Affymetrix Rat Gene 1.0 ST Array 10900167 - -
          Affymetrix Rat Gene 1.0 ST Array 10900162 - -
          Affymetrix Rat Gene 1.0 ST Array 10900168 - -
          Affymetrix Rat Gene 1.0 ST Array 10900179 - -
          Affymetrix Rat Gene 1.0 ST Array 10900166 - -
          Affymetrix Rat Gene 1.0 ST Array 10900182 - -
          Affymetrix Rat Gene 1.0 ST Array 10900164 - -
          Affymetrix Rat Gene 1.0 ST Array 10900163 - -
          Affymetrix Rat Gene 1.0 ST Array 10900175 - -
          Affymetrix Rat Gene 1.1 ST Array 10900161 - -
          Rat Genome 230 2.0 Array 1374861_PM_at - -
          Rat Genome 230 2.0 Array 1374861_at - -
          Affymetrix HT Rat Focus Array 1374861_at - -
          Affymetrix HT Rat Focus Array 1374861_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700016240.rn.1 TC0700002099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016241.rn.1 TC0700002099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016248.rn.1 TC0700002099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016238.rn.1 TC0700002099.rn.1 RTA TC
          Rat Expression Set 230 1374861_at - -
          Rat Expression Set 230 1374861_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10900161 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21850395 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361447 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline