Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536049_g1
          See other TGIF2 GE Assays ›
          Gene Symbol:
          Tgif2
          Gene Name:
          TGFB-induced factor homeobox 2
          Gene Aliases:
          RGD1560115, RGD1564927, Tgif2l1
          Chromosome Location:
          Chr.3: 152906960 - 152924380 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536049_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgif2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134983.1 NP_001128455.1 2 - 3 411 63
          XM_006235424.2 XP_006235486.1 1 - 2 228 63
          XM_006235425.2 XP_006235487.1 2 - 3 2300 63
          XM_006235426.2 XP_006235488.1 2 - 3 475 63
          XM_006235427.2 XP_006235489.1 2 - 3 497 63
          GenBank mRNA BC168719.1 - 2 - 3 411 8364226 63
          FQ235168.1 - 2 - 3 349 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of gastrulation
          nodal signaling pathway
          positive regulation of neuron differentiation
          retina development in camera-type eye
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1026139 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P044420 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P104382 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P026873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266115 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67494 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026873 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044420 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104382 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1026139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104382 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026873 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1026139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044420 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P026873 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1026139 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P044420 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P104382 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841590 - -
          Affymetrix Rat Gene 1.0 ST Array 10841588 - -
          Affymetrix Rat Gene 1.0 ST Array 10841591 - -
          Affymetrix Rat Gene 1.0 ST Array 10795417 - -
          Affymetrix Rat Gene 1.0 ST Array 10795416 - -
          Affymetrix Rat Gene 1.0 ST Array 10841587 - -
          Affymetrix Rat Gene 1.1 ST Array 10841586 - -
          Affymetrix Rat Gene 1.1 ST Array 10841589 - -
          Affymetrix Rat Gene 1.1 ST Array 10795415 - -
          Rat Genome 230 2.0 Array 1390681_PM_at - -
          Rat Genome 230 2.0 Array 1390681_at - -
          Affymetrix HT Rat Focus Array 1390681_PM_at - -
          Affymetrix HT Rat Focus Array 1390681_at - -
          Affymetrix Rat Clariom S Assay PSR0300015980.rn.1 TC0300002160.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300015979.rn.1 TC0300002160.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300015981.rn.1 TC0300002160.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300015982.rn.1 TC0300002160.rn.1 RTA TC
          Rat Expression Set 230 1390681_PM_at - -
          Rat Expression Set 230 1390681_at - -
          Affymetrix Rat Gene 1.0 ST Array 10841589 - -
          Affymetrix Rat Gene 1.0 ST Array 10841586 - -
          Affymetrix Rat Gene 1.0 ST Array 10795415 - -
          Rat Genome U34 Set rc_AA964074_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21956202 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372777 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650739 - -
          RatRef-12 Expression BeadChip ILMN_51433 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline