Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536120_m1
          See other SNPH GE Assays ›
          Gene Symbol:
          Snph
          Gene Name:
          syntaphilin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 147102394 - 147143576 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536120_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snph
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106525.3 NP_001099995.1 5 - 6 632 71
          XM_006235223.2 XP_006235285.1 3 - 4 624 71
          XM_006235224.2 XP_006235286.1 3 - 4 456 71
          XM_006235225.2 XP_006235287.1 2 - 3 348 71
          XM_006235226.2 XP_006235288.1 3 - 4 461 71
          GenBank mRNA BC168917.1 - 5 - 6 632 7132786 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004449.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          brain development
          neuron differentiation
          protein binding
          syntaxin-1 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P229581 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P144278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358535 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26237 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158499 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10149 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144278 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P229581 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P229581 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144278 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P144278 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P229581 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P229581 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92478 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10442 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10850741 - -
          Affymetrix Rat Gene 1.0 ST Array 10850742 - -
          Affymetrix Rat Gene 1.0 ST Array 10850735 - -
          Affymetrix Rat Gene 1.0 ST Array 10850733 - -
          Affymetrix Rat Gene 1.0 ST Array 10850740 - -
          Affymetrix Rat Gene 1.0 ST Array 10850734 - -
          Affymetrix Rat Gene 1.0 ST Array 10850736 - -
          Affymetrix Rat Gene 1.1 ST Array 10850732 - -
          Rat Genome 230 2.0 Array 1390540_PM_at - -
          Rat Genome 230 2.0 Array 1381627_PM_at - -
          Rat Genome 230 2.0 Array 1390540_at - -
          Rat Genome 230 2.0 Array 1381627_at - -
          Affymetrix Rat Clariom S Assay PSR0300034167.rn.1 TC0300004449.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034169.rn.1 TC0300004449.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034170.rn.1 TC0300004449.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034171.rn.1 TC0300004449.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034173.rn.1 TC0300004449.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034174.rn.1 TC0300004449.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034172.rn.1 TC0300004449.rn.1 RTA TC
          Rat Expression Set 230 1381627_at - -
          Rat Expression Set 230 1381627_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10850732 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21967564 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373272 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline