Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536695_m1
          See other GPR114 GE Assays ›
          Gene Symbol:
          Gpr114
          Gene Name:
          G protein-coupled receptor 114
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 10491449 - 10515076 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536695_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr114
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107410.1 NP_001100880.1 10 - 11 1269 90
          XM_006255101.2 XP_006255163.1 11 - 12 1310 90
          XM_008772297.1 XP_008770519.1 11 - 12 1295 90
          XM_008772298.1 XP_008770520.1 11 - 12 1224 90
          XM_008772299.1 XP_008770521.1 11 - 12 1217 90
          XM_008772300.1 XP_008770522.1 10 - 11 1406 90
          XM_008772301.1 XP_008770523.1 11 - 12 1334 90
          XM_008772302.1 XP_008770524.1 11 - 12 1256 90
          XM_008772303.1 XP_008770525.1 11 - 12 1288 90
          XM_008772304.1 XP_008770526.1 11 - 12 1595 90
          XM_008772305.1 XP_008770527.1 11 - 12 1256 90
          XM_008772306.1 XP_008770528.1 11 - 12 1329 90
          XM_008772307.1 XP_008770529.1 11 - 12 1326 90
          XM_008772308.1 XP_008770530.1 11 - 12 1329 90
          XM_008772309.1 XP_008770531.1 11 - 12 1305 90
          XM_008772310.1 XP_008770532.1 11 - 12 1283 90
          XM_008772311.1 XP_008770533.1 11 - 12 1248 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000986.rn.1

          Set Membership:

          Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157450 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505135 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148806 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157446 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157450 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157450 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157446 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157450 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157446 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P220301 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10809238 - -
          Affymetrix Rat Gene 1.0 ST Array 10809236 - -
          Affymetrix Rat Gene 1.0 ST Array 10809234 - -
          Affymetrix Rat Gene 1.0 ST Array 10809233 - -
          Affymetrix Rat Gene 1.0 ST Array 10809235 - -
          Affymetrix Rat Gene 1.0 ST Array 10809237 - -
          Affymetrix Rat Gene 1.0 ST Array 10809239 - -
          Affymetrix Rat Gene 1.1 ST Array 10809232 - -
          Rat Genome 230 2.0 Array 1391690_PM_at - -
          Rat Genome 230 2.0 Array 1391690_at - -
          Affymetrix HT Rat Focus Array 1391690_at - -
          Affymetrix HT Rat Focus Array 1391690_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900007837.rn.1 TC1900000986.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007839.rn.1 TC1900000986.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007841.rn.1 TC1900000986.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007838.rn.1 TC1900000986.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007842.rn.1 TC1900000986.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007840.rn.1 TC1900000986.rn.1 RTA TC
          Rat Expression Set 230 1391690_at - -
          Rat Expression Set 230 1391690_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10809232 - -
          Rat Genome U34 Set rc_AI171008_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21837915 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364429 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline