Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01537293_m1
          See other CLPTM1L GE Assays ›
          Gene Symbol:
          Clptm1l
          Gene Name:
          CLPTM1-like
          Gene Aliases:
          RGD1307896
          Chromosome Location:
          Chr.1: 32281207 - 32297195 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01537293_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clptm1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108240.1 NP_001101710.1 16 - 17 1728 73
          XM_006227791.2 XP_006227853.1 16 - 17 1730 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004278.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          apoptotic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628119 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115183 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5297 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082381 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082381 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10662 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58243 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10717549 - -
          Affymetrix Rat Gene 1.0 ST Array 10717553 - -
          Affymetrix Rat Gene 1.0 ST Array 10717542 - -
          Affymetrix Rat Gene 1.0 ST Array 10717544 - -
          Affymetrix Rat Gene 1.0 ST Array 10717541 - -
          Affymetrix Rat Gene 1.0 ST Array 10717539 - -
          Affymetrix Rat Gene 1.0 ST Array 10717548 - -
          Affymetrix Rat Gene 1.0 ST Array 10717538 - -
          Affymetrix Rat Gene 1.0 ST Array 10717546 - -
          Affymetrix Rat Gene 1.0 ST Array 10717540 - -
          Affymetrix Rat Gene 1.0 ST Array 10717537 - -
          Affymetrix Rat Gene 1.0 ST Array 10717545 - -
          Affymetrix Rat Gene 1.0 ST Array 10717543 - -
          Affymetrix Rat Gene 1.0 ST Array 10717547 - -
          Affymetrix Rat Gene 1.0 ST Array 10717550 - -
          Affymetrix Rat Gene 1.0 ST Array 10717551 - -
          Affymetrix Rat Gene 1.0 ST Array 10717552 - -
          Affymetrix Rat Gene 1.1 ST Array 10717536 - -
          Rat Genome 230 2.0 Array 1388861_PM_at - -
          Rat Genome 230 2.0 Array 1388861_at - -
          Affymetrix HT Rat Focus Array 1388861_at - -
          Affymetrix HT Rat Focus Array 1388861_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100034230.rn.1 TC0100004278.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100034232.rn.1 TC0100004278.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100034229.rn.1 TC0100004278.rn.1 RTA TC
          Rat Expression Set 230 1388861_at - -
          Rat Expression Set 230 1388861_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10717536 - -
          Rat Genome U34 Set rc_AI013387_at - -
          Rat Genome U34 Set rc_AI102348_at - -
          Rat Genome U34 Set rc_AI101462_at - -
          Rat Genome U34 Set rc_AI101462_g_at - -
          Rat Genome U34 Set rc_AI178551_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20890005 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350367 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline