Search
Search

Fastkd3| Gene Symbol: Fastkd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082574.1 | NP_001076043.1 | 5 - 6 | 1801 | 72 | |
| XM_006227809.2 | XP_006227871.1 | 5 - 6 | 2573 | 72 | ||
| XM_006227811.2 | XP_006227873.1 | 5 - 6 | 2014 | 72 | ||
| XM_008758678.1 | XP_008756900.1 | 4 - 5 | 2970 | 72 | ||
| XM_008758679.1 | XP_008756901.1 | 5 - 6 | 2877 | 72 | ||
| GenBank mRNA | BC079475.1 | - | 4 - 5 | 1559 | 72 | |
| BC161806.1 | - | 3 - 4 | 1597 | 7111194 | 72 | |
| FQ214620.1 | - | 4 - 5 | 354 | 72 | ||
Set Membership: |
|