Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01544829_m1
          See other COL7A1 GE Assays ›
          Gene Symbol:
          Col7a1
          Gene Name:
          collagen, type VII, alpha 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 117694441 - 117726844 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01544829_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Col7a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106858.1 NP_001100328.1 113 - 114 8375 60
          XM_006243759.2 XP_006243821.1 114 - 115 8439 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001563.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          endodermal cell differentiation
          molecular_function
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P137075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312068 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51263 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85321 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171425 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146779 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137075 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137075 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P137075 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P295563 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43137 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88822 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10913434 - -
          Affymetrix Rat Gene 1.0 ST Array 10913416 - -
          Affymetrix Rat Gene 1.0 ST Array 10913439 - -
          Affymetrix Rat Gene 1.0 ST Array 10913427 - -
          Affymetrix Rat Gene 1.0 ST Array 10913393 - -
          Affymetrix Rat Gene 1.0 ST Array 10913436 - -
          Affymetrix Rat Gene 1.0 ST Array 10913429 - -
          Affymetrix Rat Gene 1.0 ST Array 10913360 - -
          Affymetrix Rat Gene 1.0 ST Array 10913394 - -
          Affymetrix Rat Gene 1.0 ST Array 10913355 - -
          Affymetrix Rat Gene 1.0 ST Array 10913356 - -
          Affymetrix Rat Gene 1.0 ST Array 10913405 - -
          Affymetrix Rat Gene 1.0 ST Array 10913413 - -
          Affymetrix Rat Gene 1.0 ST Array 10913457 - -
          Affymetrix Rat Gene 1.0 ST Array 10913359 - -
          Affymetrix Rat Gene 1.0 ST Array 10913389 - -
          Affymetrix Rat Gene 1.0 ST Array 10913375 - -
          Affymetrix Rat Gene 1.0 ST Array 10913357 - -
          Affymetrix Rat Gene 1.0 ST Array 10913449 - -
          Affymetrix Rat Gene 1.0 ST Array 10913404 - -
          Affymetrix Rat Gene 1.0 ST Array 10913381 - -
          Affymetrix Rat Gene 1.0 ST Array 10913406 - -
          Affymetrix Rat Gene 1.0 ST Array 10913383 - -
          Affymetrix Rat Gene 1.0 ST Array 10913452 - -
          Affymetrix Rat Gene 1.0 ST Array 10913466 - -
          Affymetrix Rat Gene 1.0 ST Array 10913379 - -
          Affymetrix Rat Gene 1.0 ST Array 10913461 - -
          Affymetrix Rat Gene 1.0 ST Array 10913380 - -
          Affymetrix Rat Gene 1.0 ST Array 10913420 - -
          Affymetrix Rat Gene 1.0 ST Array 10913367 - -
          Affymetrix Rat Gene 1.0 ST Array 10913382 - -
          Affymetrix Rat Gene 1.0 ST Array 10913431 - -
          Affymetrix Rat Gene 1.0 ST Array 10913456 - -
          Affymetrix Rat Gene 1.0 ST Array 10913409 - -
          Affymetrix Rat Gene 1.0 ST Array 10913399 - -
          Affymetrix Rat Gene 1.0 ST Array 10913418 - -
          Affymetrix Rat Gene 1.0 ST Array 10913423 - -
          Affymetrix Rat Gene 1.0 ST Array 10913442 - -
          Affymetrix Rat Gene 1.0 ST Array 10913435 - -
          Affymetrix Rat Gene 1.0 ST Array 10913414 - -
          Affymetrix Rat Gene 1.0 ST Array 10913430 - -
          Affymetrix Rat Gene 1.0 ST Array 10913407 - -
          Affymetrix Rat Gene 1.0 ST Array 10913433 - -
          Affymetrix Rat Gene 1.0 ST Array 10913368 - -
          Affymetrix Rat Gene 1.0 ST Array 10913412 - -
          Affymetrix Rat Gene 1.0 ST Array 10913369 - -
          Affymetrix Rat Gene 1.0 ST Array 10913364 - -
          Affymetrix Rat Gene 1.0 ST Array 10913376 - -
          Affymetrix Rat Gene 1.0 ST Array 10913454 - -
          Affymetrix Rat Gene 1.0 ST Array 10913446 - -
          Affymetrix Rat Gene 1.0 ST Array 10913374 - -
          Affymetrix Rat Gene 1.0 ST Array 10913362 - -
          Affymetrix Rat Gene 1.0 ST Array 10913448 - -
          Affymetrix Rat Gene 1.0 ST Array 10913370 - -
          Affymetrix Rat Gene 1.0 ST Array 10913451 - -
          Affymetrix Rat Gene 1.0 ST Array 10913422 - -
          Affymetrix Rat Gene 1.0 ST Array 10913361 - -
          Affymetrix Rat Gene 1.0 ST Array 10913445 - -
          Affymetrix Rat Gene 1.0 ST Array 10913453 - -
          Affymetrix Rat Gene 1.0 ST Array 10913426 - -
          Affymetrix Rat Gene 1.0 ST Array 10913424 - -
          Affymetrix Rat Gene 1.0 ST Array 10913447 - -
          Affymetrix Rat Gene 1.0 ST Array 10913460 - -
          Affymetrix Rat Gene 1.0 ST Array 10913450 - -
          Affymetrix Rat Gene 1.0 ST Array 10913386 - -
          Affymetrix Rat Gene 1.0 ST Array 10913464 - -
          Affymetrix Rat Gene 1.0 ST Array 10913408 - -
          Affymetrix Rat Gene 1.0 ST Array 10913415 - -
          Affymetrix Rat Gene 1.0 ST Array 10913387 - -
          Affymetrix Rat Gene 1.0 ST Array 10913458 - -
          Affymetrix Rat Gene 1.0 ST Array 10913465 - -
          Affymetrix Rat Gene 1.0 ST Array 10913363 - -
          Affymetrix Rat Gene 1.0 ST Array 10913432 - -
          Affymetrix Rat Gene 1.0 ST Array 10913425 - -
          Affymetrix Rat Gene 1.0 ST Array 10913410 - -
          Affymetrix Rat Gene 1.0 ST Array 10913365 - -
          Affymetrix Rat Gene 1.0 ST Array 10913443 - -
          Affymetrix Rat Gene 1.0 ST Array 10913459 - -
          Affymetrix Rat Gene 1.0 ST Array 10913378 - -
          Affymetrix Rat Gene 1.0 ST Array 10913397 - -
          Affymetrix Rat Gene 1.0 ST Array 10913372 - -
          Affymetrix Rat Gene 1.0 ST Array 10913463 - -
          Affymetrix Rat Gene 1.0 ST Array 10913419 - -
          Affymetrix Rat Gene 1.0 ST Array 10913401 - -
          Affymetrix Rat Gene 1.0 ST Array 10913384 - -
          Affymetrix Rat Gene 1.0 ST Array 10913417 - -
          Affymetrix Rat Gene 1.0 ST Array 10913438 - -
          Affymetrix Rat Gene 1.0 ST Array 10913444 - -
          Affymetrix Rat Gene 1.0 ST Array 10913441 - -
          Affymetrix Rat Gene 1.0 ST Array 10913396 - -
          Affymetrix Rat Gene 1.0 ST Array 10913385 - -
          Affymetrix Rat Gene 1.0 ST Array 10913437 - -
          Affymetrix Rat Gene 1.0 ST Array 10913411 - -
          Affymetrix Rat Gene 1.0 ST Array 10913358 - -
          Affymetrix Rat Gene 1.0 ST Array 10913366 - -
          Affymetrix Rat Gene 1.0 ST Array 10913421 - -
          Affymetrix Rat Gene 1.0 ST Array 10913402 - -
          Affymetrix Rat Gene 1.0 ST Array 10913377 - -
          Affymetrix Rat Gene 1.0 ST Array 10913400 - -
          Affymetrix Rat Gene 1.0 ST Array 10913395 - -
          Affymetrix Rat Gene 1.0 ST Array 10913440 - -
          Affymetrix Rat Gene 1.0 ST Array 10913388 - -
          Affymetrix Rat Gene 1.0 ST Array 10913391 - -
          Affymetrix Rat Gene 1.0 ST Array 10913398 - -
          Affymetrix Rat Gene 1.0 ST Array 10913428 - -
          Affymetrix Rat Gene 1.0 ST Array 10913462 - -
          Affymetrix Rat Gene 1.0 ST Array 10913373 - -
          Affymetrix Rat Gene 1.0 ST Array 10913390 - -
          Affymetrix Rat Gene 1.0 ST Array 10913392 - -
          Affymetrix Rat Gene 1.0 ST Array 10913403 - -
          Affymetrix Rat Gene 1.1 ST Array 10913354 - -
          Rat Genome 230 2.0 Array 1374226_PM_at - -
          Rat Genome 230 2.0 Array 1374226_at - -
          Affymetrix HT Rat Focus Array 1374226_PM_at - -
          Affymetrix HT Rat Focus Array 1374226_at - -
          Affymetrix Rat Clariom S Assay PSR0800012078.rn.1 TC0800001563.rn.1 RTA TC
          Rat Expression Set 230 1374226_at - -
          Rat Expression Set 230 1374226_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10913354 - -
          Rat Genome U34 Set rc_AI171548_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21321451 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038930 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline