Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01638093_g1
          See other ASIC1 GE Assays ›
          Gene Symbol:
          Asic1
          Gene Name:
          acid-sensing (proton-gated) ion channel 1
          Gene Aliases:
          Accn2
          Chromosome Location:
          Chr.7: 141325657 - 141354937 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01638093_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Asic1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024154.2 NP_077068.1 7 - 8 1164 66
          XM_006257378.2 XP_006257440.1 5 - 6 1489 66
          XM_006257381.2 XP_006257443.1 7 - 8 1449 66
          XM_008765793.1 XP_008764015.1 5 - 6 1493 66
          XM_008765794.1 XP_008764016.1 7 - 8 1452 66
          XM_008765795.1 XP_008764017.1 7 - 8 1105 66
          XM_008765796.1 XP_008764018.1 7 - 8 1449 66
          GenBank mRNA AB049451.1 - 5 - 6 755 66
          AJ006519.1 - 5 - 6 1210 66
          AJ309926.1 - 5 - 6 1214 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001821.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Asic1

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          calcium ion transport
          memory
          associative learning
          response to acidic pH
          sensory perception of pain
          ion transmembrane transport
          sodium ion transmembrane transport
          regulation of membrane potential
          negative regulation of neurotransmitter secretion
          sensory perception of sour taste
          protein homotrimerization
          calcium ion transmembrane transport
          cellular response to pH
          cation transmembrane transport
          ion channel activity
          cation channel activity
          ion gated channel activity
          acid-sensing ion channel activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P109575 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11413 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103902 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P109575 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011050 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P109575 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011050 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P109575 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011050 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12408 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P109575 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P109573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65741 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83290 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41375 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10899171 - -
          Affymetrix Rat Gene 1.0 ST Array 10899158 - -
          Affymetrix Rat Gene 1.0 ST Array 10899164 - -
          Affymetrix Rat Gene 1.0 ST Array 10899160 - -
          Affymetrix Rat Gene 1.0 ST Array 10899159 - -
          Affymetrix Rat Gene 1.0 ST Array 10899173 - -
          Affymetrix Rat Gene 1.0 ST Array 10899163 - -
          Affymetrix Rat Gene 1.0 ST Array 10899172 - -
          Affymetrix Rat Gene 1.0 ST Array 10899166 - -
          Affymetrix Rat Gene 1.0 ST Array 10899161 - -
          Affymetrix Rat Gene 1.0 ST Array 10899167 - -
          Affymetrix Rat Gene 1.0 ST Array 10899162 - -
          Affymetrix Rat Gene 1.0 ST Array 10899168 - -
          Affymetrix Rat Gene 1.0 ST Array 10899165 - -
          Affymetrix Rat Gene 1.0 ST Array 10899170 - -
          Affymetrix Rat Gene 1.1 ST Array 10899157 - -
          Rat Genome 230 2.0 Array 1387555_PM_at - -
          Rat Genome 230 2.0 Array 1371045_at - -
          Rat Genome 230 2.0 Array 1388078_a_at - -
          Rat Genome 230 2.0 Array 1371045_PM_at - -
          Rat Genome 230 2.0 Array 1388078_PM_a_at - -
          Rat Genome 230 2.0 Array 1392135_at - -
          Rat Genome 230 2.0 Array 1387555_at - -
          Rat Genome 230 2.0 Array 1392135_PM_at - -
          Affymetrix HT Rat Focus Array 1388078_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387555_at - -
          Affymetrix HT Rat Focus Array 1388078_a_at - -
          Affymetrix HT Rat Focus Array 1392135_PM_at - -
          Affymetrix HT Rat Focus Array 1392135_at - -
          Affymetrix HT Rat Focus Array 1371045_at - -
          Affymetrix HT Rat Focus Array 1371045_PM_at - -
          Affymetrix HT Rat Focus Array 1387555_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700014042.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014041.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014062.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014058.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014061.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014045.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014052.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014044.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014057.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014055.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014051.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014043.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014056.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014060.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014039.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014059.rn.1 TC0700001821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014063.rn.1 TC0700001821.rn.1 RTA TC
          Rat Expression Set 230 1387555_PM_at - -
          Rat Expression Set 230 1371045_PM_at - -
          Rat Expression Set 230 1388078_PM_a_at - -
          Rat Expression Set 230 1388078_a_at - -
          Rat Expression Set 230 1387555_at - -
          Rat Expression Set 230 1371045_at - -
          Rat Expression Set 230 1392135_PM_at - -
          Rat Expression Set 230 1392135_at - -
          Affymetrix Rat Gene 1.0 ST Array 10899157 - -
          Rat Genome U34 Set AJ006519_at - -
          Rat Genome U34 Set AJ006519_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354195 - -
          RatRef-12 Expression BeadChip ILMN_48926 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline