Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01638706_m1
          See other ENPP1 GE Assays ›
          Gene Symbol:
          Enpp1
          Gene Name:
          ectonucleotide pyrophosphatase/phosphodiesterase 1
          Gene Aliases:
          Npps, Pc1
          Chromosome Location:
          Chr.1: 21748214 - 21813205 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01638706_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Enpp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053535.1 NP_445987.1 14 - 15 1389 87
          XM_006227692.2 XP_006227754.1 14 - 15 1411 87
          GenBank mRNA AF320054.1 - 14 - 15 1389 87
          AF340185.1 - 14 - 15 1389 87
          AF340186.1 - 14 - 15 1389 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000233.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Rat Phosphodiesterase 1700 Inventoried Rat Phosphodiesterase Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Enpp1

          Gene Ontology Categories:

          Function(s) Process(es)

          generation of precursor metabolites and energy
          phosphate-containing compound metabolic process
          receptor-mediated endocytosis
          immune response
          biological_process
          nucleoside triphosphate catabolic process
          negative regulation of ossification
          negative regulation of cell growth
          inorganic diphosphate transport
          cellular phosphate ion homeostasis
          sequestering of triglyceride
          biomineral tissue development
          negative regulation of protein autophosphorylation
          cellular response to insulin stimulus
          negative regulation of fat cell differentiation
          negative regulation of glycogen biosynthetic process
          negative regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          bone remodeling
          3'-phosphoadenosine 5'-phosphosulfate metabolic process
          nucleic acid phosphodiester bond hydrolysis
          molecular_function
          nucleic acid binding
          phosphodiesterase I activity
          nucleotide diphosphatase activity
          scavenger receptor activity
          insulin receptor binding
          calcium ion binding
          ATP binding
          zinc ion binding
          polysaccharide binding
          NADH pyrophosphatase activity
          protein homodimerization activity
          metal ion binding
          nucleoside-triphosphate diphosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312266 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141567 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170267 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18837 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12841 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12841 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12841 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12841 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61141 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90823 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75618 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98949 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104939 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702265 - -
          Affymetrix Rat Gene 1.0 ST Array 10702268 - -
          Affymetrix Rat Gene 1.0 ST Array 10702264 - -
          Affymetrix Rat Gene 1.0 ST Array 10702255 - -
          Affymetrix Rat Gene 1.0 ST Array 10702258 - -
          Affymetrix Rat Gene 1.0 ST Array 10702261 - -
          Affymetrix Rat Gene 1.0 ST Array 10702257 - -
          Affymetrix Rat Gene 1.0 ST Array 10702260 - -
          Affymetrix Rat Gene 1.0 ST Array 10702270 - -
          Affymetrix Rat Gene 1.0 ST Array 10702252 - -
          Affymetrix Rat Gene 1.0 ST Array 10702271 - -
          Affymetrix Rat Gene 1.0 ST Array 10702275 - -
          Affymetrix Rat Gene 1.0 ST Array 10702266 - -
          Affymetrix Rat Gene 1.0 ST Array 10702272 - -
          Affymetrix Rat Gene 1.0 ST Array 10702273 - -
          Affymetrix Rat Gene 1.0 ST Array 10702269 - -
          Affymetrix Rat Gene 1.0 ST Array 10702256 - -
          Affymetrix Rat Gene 1.0 ST Array 10702251 - -
          Affymetrix Rat Gene 1.0 ST Array 10702253 - -
          Affymetrix Rat Gene 1.0 ST Array 10702262 - -
          Affymetrix Rat Gene 1.0 ST Array 10702274 - -
          Affymetrix Rat Gene 1.0 ST Array 10702267 - -
          Affymetrix Rat Gene 1.0 ST Array 10702254 - -
          Affymetrix Rat Gene 1.0 ST Array 10702259 - -
          Affymetrix Rat Gene 1.0 ST Array 10702263 - -
          Affymetrix Rat Gene 1.1 ST Array 10702250 - -
          Rat Genome 230 2.0 Array 1370047_PM_at - -
          Rat Genome 230 2.0 Array 1370047_at - -
          Affymetrix HT Rat Focus Array 1370047_PM_at - -
          Affymetrix HT Rat Focus Array 1370047_at - -
          Affymetrix Rat Clariom S Assay PSR0100001339.rn.1 TC0100000233.rn.1 RTA TC
          Rat Expression Set 230 1370047_at - -
          Rat Expression Set 230 1370047_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10702250 - -
          Rat Genome U34 Set AB017596_at - -
          Rat Genome U34 Set rc_AA817968_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22030410 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365255 - -
          RatRef-12 Expression BeadChip ILMN_62994 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline