Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01639345_m1
          See other PDE5A GE Assays ›
          Gene Symbol:
          Pde5a
          Gene Name:
          phosphodiesterase 5A, cGMP-specific
          Gene Aliases:
          PDE5A2
          Chromosome Location:
          Chr.2: 226899409 - 227044916 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01639345_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pde5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133584.1 NP_598268.1 15 - 16 2435 93
          XM_006233260.2 XP_006233322.1 15 - 16 2363 93
          GenBank mRNA D89093.1 - 15 - 16 2435 93
          U76032.1 - 6 - 7 551 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002121.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Rat Phosphodiesterase 1700 Inventoried Rat Phosphodiesterase Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pde5a

          Panther Classification:

          Molecular Function -

          phosphodiesterase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          regulation of the force of heart contraction
          positive regulation of chronic inflammatory response
          signal transduction
          nervous system development
          short-term memory
          positive regulation of cardiac muscle hypertrophy
          regulation of cGMP metabolic process
          response to lipopolysaccharide
          response to testosterone
          negative regulation of T cell proliferation
          vasodilation
          positive regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of vasoconstriction
          cGMP metabolic process
          cGMP catabolic process
          negative regulation of cardiac muscle contraction
          relaxation of cardiac muscle
          positive regulation of oocyte development
          cyclic-nucleotide phosphodiesterase activity
          3',5'-cyclic-nucleotide phosphodiesterase activity
          cGMP binding
          metal ion binding
          3',5'-cyclic-GMP phosphodiesterase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178297 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114416 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28805 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76992 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72263 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77790 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53231 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8296 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47963 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62117 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36434 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81577 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15466 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15466 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1967 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18870 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89718 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93185 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71927 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11490 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54058 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66964 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97458 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818845 - -
          Affymetrix Rat Gene 1.0 ST Array 10818840 - -
          Affymetrix Rat Gene 1.0 ST Array 10818830 - -
          Affymetrix Rat Gene 1.0 ST Array 10818842 - -
          Affymetrix Rat Gene 1.0 ST Array 10818837 - -
          Affymetrix Rat Gene 1.0 ST Array 10818834 - -
          Affymetrix Rat Gene 1.0 ST Array 10818827 - -
          Affymetrix Rat Gene 1.0 ST Array 10818828 - -
          Affymetrix Rat Gene 1.0 ST Array 10818836 - -
          Affymetrix Rat Gene 1.0 ST Array 10818835 - -
          Affymetrix Rat Gene 1.0 ST Array 10818829 - -
          Affymetrix Rat Gene 1.0 ST Array 10818832 - -
          Affymetrix Rat Gene 1.0 ST Array 10818841 - -
          Affymetrix Rat Gene 1.0 ST Array 10818844 - -
          Affymetrix Rat Gene 1.0 ST Array 10818838 - -
          Affymetrix Rat Gene 1.0 ST Array 10818843 - -
          Affymetrix Rat Gene 1.0 ST Array 10818825 - -
          Affymetrix Rat Gene 1.0 ST Array 10818831 - -
          Affymetrix Rat Gene 1.0 ST Array 10818826 - -
          Affymetrix Rat Gene 1.1 ST Array 10818823 - -
          Rat Genome 230 2.0 Array 1369229_at - -
          Rat Genome 230 2.0 Array 1369229_PM_at - -
          Affymetrix HT Rat Focus Array 1369229_at - -
          Affymetrix HT Rat Focus Array 1369229_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200014693.rn.1 TC0200002121.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200014694.rn.1 TC0200002121.rn.1 RTA TC
          Rat Expression Set 230 1369229_PM_at - -
          Rat Expression Set 230 1369229_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818823 - -
          Rat Genome U34 Set D89093_at - -
          Rat Genome U34 Set U76032_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22223800 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367350 - -
          RatRef-12 Expression BeadChip ILMN_60307 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline