Search
Search
| Gene Symbol: Sdhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198788.2 | NP_942083.1 | 3 - 4 | 322 | 88 | |
| GenBank mRNA | BC059160.1 | - | 3 - 4 | 327 | 6887444 | 88 |
| FQ209438.1 | - | 3 - 4 | 323 | 88 | ||
| FQ212791.1 | - | 3 - 4 | 325 | 88 | ||
| FQ212873.1 | - | 3 - 4 | 325 | 88 | ||
| FQ212985.1 | - | 3 - 4 | 324 | 88 | ||
| FQ213509.1 | - | 3 - 4 | 324 | 88 | ||
| FQ214315.1 | - | 3 - 4 | 352 | 88 | ||
| FQ215255.1 | - | 3 - 4 | 329 | 88 | ||
| FQ215355.1 | - | 3 - 4 | 323 | 88 | ||
| FQ215505.1 | - | 3 - 4 | 324 | 88 | ||
| FQ215951.1 | - | 3 - 4 | 324 | 88 | ||
| FQ215986.1 | - | 3 - 4 | 323 | 88 | ||
| FQ216027.1 | - | 3 - 4 | 324 | 88 | ||
| FQ216314.1 | - | 3 - 4 | 330 | 88 | ||
| FQ216503.1 | - | 3 - 4 | 327 | 88 | ||
| FQ220862.1 | - | 3 - 4 | 323 | 88 | ||
| FQ230166.1 | - | 3 - 4 | 316 | 88 | ||
Set Membership: |
|