Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01746552_m1
          See other CMC1 GE Assays ›
          Gene Symbol:
          Cmc1
          Gene Name:
          COX assembly mitochondrial protein 1
          Gene Aliases:
          RGD1305283
          Chromosome Location:
          Chr.8: 126437345 - 126495347 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01746552_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cmc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135259.2 NP_001128731.1 2 - 3 260 63
          NM_001199225.1 NP_001186154.1 2 - 3 260 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003521.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P057543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745753 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31786 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 573 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1410 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057543 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P057543 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P057548 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P457447 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10920800 - -
          Affymetrix Rat Gene 1.0 ST Array 10920799 - -
          Affymetrix Rat Gene 1.1 ST Array 10920797 - -
          Rat Genome 230 2.0 Array 1391776_at - -
          Rat Genome 230 2.0 Array 1391776_PM_at - -
          Affymetrix HT Rat Focus Array 1391776_at - -
          Affymetrix HT Rat Focus Array 1391776_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800028640.rn.1 TC0800003521.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800028639.rn.1 TC0800003521.rn.1 RTA TC
          Rat Expression Set 230 1391776_PM_at - -
          Rat Expression Set 230 1391776_at - -
          Affymetrix Rat Gene 1.0 ST Array 10920797 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355117 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline