Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749686_m1
          See other TCEA3 GE Assays ›
          Gene Symbol:
          Tcea3
          Gene Name:
          transcription elongation factor A (SII), 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 154598765 - 154634687 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749686_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcea3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001015008.1 NP_001015008.1 5 - 6 530 109
          GenBank mRNA BC091180.1 - 5 - 6 530 7325833 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001702.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcea3

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          translational elongation
          biological_process
          regulation of DNA-templated transcription, elongation
          molecular_function
          DNA binding
          translation elongation factor activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P084018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120331 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89695 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084018 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084018 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P084018 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1032752 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96201 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33787 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872987 - -
          Affymetrix Rat Gene 1.0 ST Array 10872988 - -
          Affymetrix Rat Gene 1.0 ST Array 10872979 - -
          Affymetrix Rat Gene 1.0 ST Array 10872984 - -
          Affymetrix Rat Gene 1.0 ST Array 10872980 - -
          Affymetrix Rat Gene 1.0 ST Array 10872985 - -
          Affymetrix Rat Gene 1.0 ST Array 10872986 - -
          Affymetrix Rat Gene 1.0 ST Array 10872990 - -
          Affymetrix Rat Gene 1.0 ST Array 10872983 - -
          Affymetrix Rat Gene 1.0 ST Array 10872981 - -
          Affymetrix Rat Gene 1.0 ST Array 10872989 - -
          Affymetrix Rat Gene 1.0 ST Array 10872982 - -
          Affymetrix Rat Gene 1.1 ST Array 10872978 - -
          Rat Genome 230 2.0 Array 1388611_at - -
          Rat Genome 230 2.0 Array 1388611_PM_at - -
          Affymetrix HT Rat Focus Array 1388611_PM_at - -
          Affymetrix HT Rat Focus Array 1388611_at - -
          Affymetrix Rat Clariom S Assay PSR0500012408.rn.1 TC0500001702.rn.1 RTA TC
          Rat Expression Set 230 1388611_PM_at - -
          Rat Expression Set 230 1388611_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872978 - -
          Rat Genome U34 Set rc_AI172272_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21745561 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361779 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline