Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01750044_m1
          See other TMEM38B GE Assays ›
          Gene Symbol:
          Tmem38b
          Gene Name:
          transmembrane protein 38B
          Gene Aliases:
          RGD1305703
          Chromosome Location:
          Chr.5: 70639156 - 70675055 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01750044_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem38b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014191.1 NP_001014213.1 3 - 4 483 82
          GenBank mRNA BC079336.1 - 3 - 4 483 7106021 82
          FQ229075.1 - 3 - 4 524 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000666.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem38b

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          potassium ion transmembrane transport
          potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P504549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105121 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107046 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P504549 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P504549 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P504549 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P795688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42195 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10869154 - -
          Affymetrix Rat Gene 1.0 ST Array 10869155 - -
          Affymetrix Rat Gene 1.0 ST Array 10869153 - -
          Affymetrix Rat Gene 1.0 ST Array 10869156 - -
          Affymetrix Rat Gene 1.0 ST Array 10869157 - -
          Affymetrix Rat Gene 1.0 ST Array 10869152 - -
          Affymetrix Rat Gene 1.1 ST Array 10869151 - -
          Rat Genome 230 2.0 Array 1376719_at - -
          Rat Genome 230 2.0 Array 1383715_at - -
          Rat Genome 230 2.0 Array 1376719_PM_at - -
          Rat Genome 230 2.0 Array 1383715_PM_at - -
          Affymetrix HT Rat Focus Array 1383715_at - -
          Affymetrix HT Rat Focus Array 1376719_PM_at - -
          Affymetrix HT Rat Focus Array 1376719_at - -
          Affymetrix HT Rat Focus Array 1383715_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500004560.rn.1 TC0500000666.rn.1 RTA TC
          Rat Expression Set 230 1376719_at - -
          Rat Expression Set 230 1376719_PM_at - -
          Rat Expression Set 230 1383715_at - -
          Rat Expression Set 230 1383715_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10869151 - -
          Rat Genome U34 Set rc_AI172189_at - -
          Rat Genome U34 Set rc_AI103376_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21077760 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline