Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753748_m1
          See other LYPD1 GE Assays ›
          Gene Symbol:
          Lypd1
          Gene Name:
          Ly6/Plaur domain containing 1
          Gene Aliases:
          Lypdc1
          Chromosome Location:
          Chr.13: 42219884 - 42263024 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753748_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lypd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007727.1 NP_001007728.1 2 - 3 490 78
          GenBank mRNA BC082032.1 - 2 - 3 490 7130639 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001316.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lypd1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P763058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142636 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66111 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89746 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99557 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P763058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P763058 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P763058 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19561 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23052 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103530 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63444 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767246 - -
          Affymetrix Rat Gene 1.0 ST Array 10767245 - -
          Affymetrix Rat Gene 1.0 ST Array 10763606 - -
          Affymetrix Rat Gene 1.0 ST Array 10767244 - -
          Affymetrix Rat Gene 1.1 ST Array 10767243 - -
          Affymetrix Rat Gene 1.1 ST Array 10763604 - -
          Affymetrix Rat Clariom S Assay PSR1300008634.rn.1 TC1300001316.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10767243 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22129155 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352757 - -
          RatRef-12 Expression BeadChip ILMN_53274 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline