Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753772_m1
          See other INHBB GE Assays ›
          Gene Symbol:
          Inhbb
          Gene Name:
          inhibin beta-B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 35436532 - 35442222 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753772_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Inhbb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080771.1 NP_542949.1 1 - 2 458 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001255.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          male gonad development
          cellular response to starvation
          response to mechanical stimulus
          positive regulation of pathway-restricted SMAD protein phosphorylation
          response to organic cyclic compound
          response to insecticide
          pituitary gland development
          cellular response to insulin stimulus
          activin receptor signaling pathway
          response to gonadotropin
          growth
          regulation of apoptotic process
          regulation of MAPK cascade
          fat cell differentiation
          negative regulation of insulin secretion
          positive regulation of follicle-stimulating hormone secretion
          negative regulation of follicle-stimulating hormone secretion
          negative regulation of hepatocyte growth factor biosynthetic process
          cell development
          oocyte development
          positive regulation of ovulation
          SMAD protein signal transduction
          cellular response to cAMP
          cellular response to interleukin-1
          cellular response to cholesterol
          cellular response to organic cyclic compound
          cellular response to thyroid hormone stimulus
          positive regulation of apoptotic signaling pathway
          cytokine activity
          transforming growth factor beta receptor binding
          hormone activity
          growth factor activity
          inhibin binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P030644 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P576822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P576822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030644 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P576822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030644 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P030644 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P576822 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P576822 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P332702 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288117 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767096 - -
          Affymetrix Rat Gene 1.1 ST Array 10767095 - -
          Rat Genome 230 2.0 Array 1377163_PM_at - -
          Rat Genome 230 2.0 Array 1377163_at - -
          Affymetrix HT Rat Focus Array 1377163_PM_at - -
          Affymetrix HT Rat Focus Array 1377163_at - -
          Affymetrix Rat Clariom S Assay PSR1300008275.rn.1 TC1300001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008276.rn.1 TC1300001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008273.rn.1 TC1300001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008278.rn.1 TC1300001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008274.rn.1 TC1300001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008272.rn.1 TC1300001255.rn.1 RTA TC
          Rat Expression Set 230 1377163_PM_at - -
          Rat Expression Set 230 1377163_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767095 - -
          Rat Genome U34 Set M32757cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21090942 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363849 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530450 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline