Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01754276_m1
          See other TPD52L2 GE Assays ›
          Gene Symbol:
          Tpd52l2
          Gene Name:
          tumor protein D52-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 176989653 - 177009425 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01754276_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tpd52l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198744.2 NP_942039.1 7 - 8 722 88
          XM_006235754.2 XP_006235816.1 8 - 9 748 88
          XM_006235757.2 XP_006235819.1 7 - 8 708 88
          XM_006235759.2 XP_006235821.1 6 - 7 672 88
          XM_006235760.2 XP_006235822.1 6 - 7 676 88
          XM_006235761.2 XP_006235823.1 5 - 6 617 88
          XM_008762499.1 XP_008760721.1 8 - 9 682 88
          XM_008762500.1 XP_008760722.1 5 - 6 551 88
          GenBank mRNA BC059167.1 - 7 - 8 723 6890828 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002603.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tpd52l2

          Gene Ontology Categories:

          Process(es)

          protein homodimerization activity
          poly(A) RNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P125496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554918 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94968 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125496 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125496 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P125496 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P418163 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P515247 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80319 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65655 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10843031 - -
          Affymetrix Rat Gene 1.0 ST Array 10843030 - -
          Affymetrix Rat Gene 1.0 ST Array 10843032 - -
          Affymetrix Rat Gene 1.0 ST Array 10843029 - -
          Affymetrix Rat Gene 1.0 ST Array 10843033 - -
          Affymetrix Rat Gene 1.0 ST Array 10843037 - -
          Affymetrix Rat Gene 1.0 ST Array 10843038 - -
          Affymetrix Rat Gene 1.0 ST Array 10843036 - -
          Affymetrix Rat Gene 1.0 ST Array 10843035 - -
          Affymetrix Rat Gene 1.1 ST Array 10843028 - -
          Rat Genome 230 2.0 Array 1372436_at - -
          Rat Genome 230 2.0 Array 1372436_PM_at - -
          Affymetrix HT Rat Focus Array 1372436_at - -
          Affymetrix HT Rat Focus Array 1372436_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300019288.rn.1 TC0300002603.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019285.rn.1 TC0300002603.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019287.rn.1 TC0300002603.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019286.rn.1 TC0300002603.rn.1 RTA TC
          Rat Expression Set 230 1372436_PM_at - -
          Rat Expression Set 230 1372436_at - -
          Affymetrix Rat Gene 1.0 ST Array 10843028 - -
          Rat Genome U34 Set rc_AI103637_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21179994 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364434 - -
          RatRef-12 Expression BeadChip ILMN_56259 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline