Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756664_m1
          See other TEX29 GE Assays ›
          Gene Symbol:
          Tex29
          Gene Name:
          testis expressed 29
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 82985834 - 83006555 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756664_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex29
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017508.1 NP_001017508.1 4 - 5 373 64
          XM_006253469.2 XP_006253531.2 3 - 4 7126 64
          XM_008771404.1 XP_008769626.1 3 - 4 7212 64
          XM_008771405.1 XP_008769627.1 2 - 3 406 64
          XM_008771406.1 XP_008769628.1 4 - 5 7101 64
          GenBank mRNA BC079268.1 - 4 - 5 373 7133664 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600002040.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tex29

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180472 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054576 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054576 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054576 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P800970 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91569 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96254 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792910 - -
          Affymetrix Rat Gene 1.0 ST Array 10792909 - -
          Affymetrix Rat Gene 1.0 ST Array 10792911 - -
          Affymetrix Rat Gene 1.0 ST Array 10792908 - -
          Affymetrix Rat Gene 1.0 ST Array 10792912 - -
          Affymetrix Rat Gene 1.0 ST Array 10792907 - -
          Affymetrix Rat Gene 1.1 ST Array 10792906 - -
          Affymetrix Rat Clariom S Assay PSR1600015209.rn.1 TC1600002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015212.rn.1 TC1600002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015215.rn.1 TC1600002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015218.rn.1 TC1600002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015221.rn.1 TC1600002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015213.rn.1 TC1600002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015219.rn.1 TC1600002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015210.rn.1 TC1600002040.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015211.rn.1 TC1600002040.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10792906 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22312207 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352454 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline