Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01757609_m1
          See other RPGRIP1L GE Assays ›
          Gene Symbol:
          Rpgrip1l
          Gene Name:
          Rpgrip1-like
          Gene Aliases:
          RGD1311099
          Chromosome Location:
          Chr.19: 17115412 - 17208055 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01757609_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpgrip1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107414.1 NP_001100884.1 10 - 11 1315 66
          XM_008772313.1 XP_008770535.1 9 - 10 1743 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000184.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          establishment of planar polarity
          kidney development
          liver development
          establishment or maintenance of cell polarity
          determination of left/right symmetry
          brain development
          biological_process
          regulation of smoothened signaling pathway
          neural tube patterning
          telencephalon development
          cerebellum development
          lateral ventricle development
          olfactory bulb development
          corpus callosum development
          limb morphogenesis
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          cilium assembly
          camera-type eye development
          nose development
          negative regulation of G-protein coupled receptor protein signaling pathway
          pericardium development
          cilium morphogenesis
          head development
          cochlea development
          molecular_function
          thromboxane A2 receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P461849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489248 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30211 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178788 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90903 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156936 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36189 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P461849 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P461849 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P461849 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461849 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9043 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23746 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15040 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806275 - -
          Affymetrix Rat Gene 1.0 ST Array 10806271 - -
          Affymetrix Rat Gene 1.0 ST Array 10806260 - -
          Affymetrix Rat Gene 1.0 ST Array 10806258 - -
          Affymetrix Rat Gene 1.0 ST Array 10806264 - -
          Affymetrix Rat Gene 1.0 ST Array 10806266 - -
          Affymetrix Rat Gene 1.0 ST Array 10806268 - -
          Affymetrix Rat Gene 1.0 ST Array 10806262 - -
          Affymetrix Rat Gene 1.0 ST Array 10806281 - -
          Affymetrix Rat Gene 1.0 ST Array 10806272 - -
          Affymetrix Rat Gene 1.0 ST Array 10806277 - -
          Affymetrix Rat Gene 1.0 ST Array 10806263 - -
          Affymetrix Rat Gene 1.0 ST Array 10806265 - -
          Affymetrix Rat Gene 1.0 ST Array 10806261 - -
          Affymetrix Rat Gene 1.0 ST Array 10806267 - -
          Affymetrix Rat Gene 1.0 ST Array 10806273 - -
          Affymetrix Rat Gene 1.0 ST Array 10806270 - -
          Affymetrix Rat Gene 1.0 ST Array 10806280 - -
          Affymetrix Rat Gene 1.0 ST Array 10806282 - -
          Affymetrix Rat Gene 1.0 ST Array 10806279 - -
          Affymetrix Rat Gene 1.0 ST Array 10806274 - -
          Affymetrix Rat Gene 1.0 ST Array 10806278 - -
          Affymetrix Rat Gene 1.0 ST Array 10806259 - -
          Affymetrix Rat Gene 1.0 ST Array 10806276 - -
          Affymetrix Rat Gene 1.0 ST Array 10806269 - -
          Affymetrix Rat Gene 1.1 ST Array 10806257 - -
          Affymetrix Rat Clariom S Assay PSR1900001436.rn.1 TC1900000184.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10806257 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22208619 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361295 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline