Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01757989_m1
          See other MAP3K4 GE Assays ›
          Gene Symbol:
          Map3k4
          Gene Name:
          mitogen activated protein kinase kinase kinase 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 48625686 - 48711642 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01757989_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Map3k4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107456.1 NP_001100926.1 23 - 24 4396 70
          XM_006227872.2 XP_006227934.1 26 - 27 4953 70
          XM_006227873.2 XP_006227935.1 25 - 26 4791 70
          XM_006227874.2 XP_006227936.1 25 - 26 4710 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000515.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Protein Kinase 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          activation of MAPK activity
          placenta development
          protein phosphorylation
          biological_process
          response to UV-C
          regulation of gene expression
          male germ-line sex determination
          intracellular signal transduction
          positive regulation of JUN kinase activity
          chorionic trophoblast cell differentiation
          positive regulation of p38MAPK cascade
          molecular_function
          protein kinase activity
          MAP kinase kinase activity
          MAP kinase kinase kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P492013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765130 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90715 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136480 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49740 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103146 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30340 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158818 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111517 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P492013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P492013 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P492013 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P492013 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66766 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90101 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10717999 - -
          Affymetrix Rat Gene 1.0 ST Array 10703095 - -
          Affymetrix Rat Gene 1.0 ST Array 10703100 - -
          Affymetrix Rat Gene 1.0 ST Array 10703093 - -
          Affymetrix Rat Gene 1.0 ST Array 10703097 - -
          Affymetrix Rat Gene 1.0 ST Array 10703094 - -
          Affymetrix Rat Gene 1.0 ST Array 10703091 - -
          Affymetrix Rat Gene 1.0 ST Array 10717997 - -
          Affymetrix Rat Gene 1.0 ST Array 10703083 - -
          Affymetrix Rat Gene 1.0 ST Array 10718000 - -
          Affymetrix Rat Gene 1.0 ST Array 10703101 - -
          Affymetrix Rat Gene 1.0 ST Array 10703087 - -
          Affymetrix Rat Gene 1.0 ST Array 10703076 - -
          Affymetrix Rat Gene 1.0 ST Array 10703098 - -
          Affymetrix Rat Gene 1.0 ST Array 10703081 - -
          Affymetrix Rat Gene 1.0 ST Array 10703082 - -
          Affymetrix Rat Gene 1.0 ST Array 10703089 - -
          Affymetrix Rat Gene 1.0 ST Array 10703079 - -
          Affymetrix Rat Gene 1.0 ST Array 10717996 - -
          Affymetrix Rat Gene 1.0 ST Array 10703086 - -
          Affymetrix Rat Gene 1.0 ST Array 10703084 - -
          Affymetrix Rat Gene 1.0 ST Array 10703090 - -
          Affymetrix Rat Gene 1.0 ST Array 10717998 - -
          Affymetrix Rat Gene 1.0 ST Array 10703085 - -
          Affymetrix Rat Gene 1.0 ST Array 10703078 - -
          Affymetrix Rat Gene 1.0 ST Array 10703088 - -
          Affymetrix Rat Gene 1.0 ST Array 10703096 - -
          Affymetrix Rat Gene 1.0 ST Array 10703099 - -
          Affymetrix Rat Gene 1.1 ST Array 10703075 - -
          Affymetrix Rat Gene 1.1 ST Array 10717995 - -
          Rat Genome 230 2.0 Array 1378791_at - -
          Rat Genome 230 2.0 Array 1378791_PM_at - -
          Affymetrix HT Rat Focus Array 1378791_PM_at - -
          Affymetrix HT Rat Focus Array 1378791_at - -
          Affymetrix Rat Clariom S Assay PSR0100003387.rn.1 TC0100000515.rn.1 RTA TC
          Rat Expression Set 230 1378791_at - -
          Rat Expression Set 230 1378791_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10703075 - -
          Rat Genome U34 Set rc_AA957257_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22021816 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373545 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0