Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01758992_m1
          See other BLZF1 GE Assays ›
          Gene Symbol:
          Blzf1
          Gene Name:
          basic leucine zipper nuclear factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 82592170 - 82607379 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01758992_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Blzf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017494.1 NP_001017494.1 3 - 4 622 97
          GenBank mRNA BC079114.1 - 3 - 4 622 7126053 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001800.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Blzf1

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          Golgi organization
          biological_process
          Golgi to plasma membrane protein transport
          protein binding
          enzyme binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P19066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155395 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19066 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19066 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P759192 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P928735 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13200 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769474 - -
          Affymetrix Rat Gene 1.0 ST Array 10769475 - -
          Affymetrix Rat Gene 1.0 ST Array 10769471 - -
          Affymetrix Rat Gene 1.0 ST Array 10769469 - -
          Affymetrix Rat Gene 1.0 ST Array 10769472 - -
          Affymetrix Rat Gene 1.0 ST Array 10769473 - -
          Affymetrix Rat Gene 1.0 ST Array 10769470 - -
          Affymetrix Rat Gene 1.1 ST Array 10769468 - -
          Rat Genome 230 2.0 Array 1392517_at - -
          Rat Genome 230 2.0 Array 1392517_PM_at - -
          Rat Genome 230 2.0 Array 1395098_PM_at - -
          Rat Genome 230 2.0 Array 1395098_at - -
          Affymetrix HT Rat Focus Array 1395098_at - -
          Affymetrix HT Rat Focus Array 1392517_at - -
          Affymetrix HT Rat Focus Array 1392517_PM_at - -
          Affymetrix HT Rat Focus Array 1395098_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300012755.rn.1 TC1300001800.rn.1 RTA TC
          Rat Expression Set 230 1395098_PM_at - -
          Rat Expression Set 230 1395098_at - -
          Rat Expression Set 230 1392517_at - -
          Rat Expression Set 230 1392517_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769468 - -
          Rat Genome U34 Set rc_AA965035_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359745 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline