Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01761189_m1
          See other CPPED1 GE Assays ›
          Gene Symbol:
          Cpped1
          Gene Name:
          calcineurin-like phosphoesterase domain containing 1
          Gene Aliases:
          RGD1306568
          Chromosome Location:
          Chr.10: 3655038 - 3774887 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01761189_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cpped1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013963.1 NP_001013985.1 3 - 4 764 61
          GenBank mRNA BC081991.1 - 3 - 4 764 7129542 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000048.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cpped1

          Gene Ontology Categories:

          Function(s) Process(es)

          dephosphorylation
          phosphoprotein phosphatase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P274762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100319 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43947 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33257 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168552 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P274762 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P274762 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P274762 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P274762 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90212 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62380 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11218 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21620 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55478 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88006 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7484 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74044 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11681 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10731423 - -
          Affymetrix Rat Gene 1.0 ST Array 10731425 - -
          Affymetrix Rat Gene 1.0 ST Array 10731427 - -
          Affymetrix Rat Gene 1.0 ST Array 10731422 - -
          Affymetrix Rat Gene 1.0 ST Array 10731424 - -
          Affymetrix Rat Gene 1.1 ST Array 10731421 - -
          Rat Genome 230 2.0 Array 1373924_at - -
          Rat Genome 230 2.0 Array 1373924_PM_at - -
          Affymetrix HT Rat Focus Array 1373924_at - -
          Affymetrix HT Rat Focus Array 1373924_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000000421.rn.1 TC1000000048.rn.1 RTA TC
          Rat Expression Set 230 1373924_at - -
          Rat Expression Set 230 1373924_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10731421 - -
          Rat Genome U34 Set rc_AI059701_at - -
          Rat Genome U34 Set rc_AI176928_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21969235 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354853 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline