Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01761388_m1
          See other PCDH18 GE Assays ›
          Gene Symbol:
          Pcdh18
          Gene Name:
          protocadherin 18
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 138819957 - 138833930 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01761388_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcdh18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100524.1 NP_001093994.1 3 - 4 3158 79
          XM_006232322.2 XP_006232384.1 3 - 4 3220 79
          GenBank mRNA BC168716.1 - 3 - 4 3158 9099515 79
          DQ863135.1 - 1 - 2 84 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003623.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          brain development
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P158123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41533 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158123 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158123 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P158123 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P454763 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823055 - -
          Affymetrix Rat Gene 1.0 ST Array 10823056 - -
          Affymetrix Rat Gene 1.0 ST Array 10823053 - -
          Affymetrix Rat Gene 1.0 ST Array 10823054 - -
          Affymetrix Rat Gene 1.1 ST Array 10823052 - -
          Rat Genome 230 2.0 Array 1384824_at - -
          Rat Genome 230 2.0 Array 1384824_PM_at - -
          Affymetrix HT Rat Focus Array 1384824_at - -
          Affymetrix HT Rat Focus Array 1384824_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200024443.rn.1 TC0200003623.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200024444.rn.1 TC0200003623.rn.1 RTA TC
          Rat Expression Set 230 1384824_at - -
          Rat Expression Set 230 1384824_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823052 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20746195 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357835 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline