Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01764746_g1
          See other GALT GE Assays ›
          Gene Symbol:
          Galt
          Gene Name:
          galactose-1-phosphate uridylyltransferase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 58144705 - 58147946 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01764746_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Galt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013089.2 NP_001013107.1 10 - 11 1134 75
          XM_006238033.2 XP_006238095.1 10 - 11 1058 75
          XM_008763615.1 XP_008761837.1 8 - 9 1022 75
          GenBank mRNA BC098756.1 - 10 - 11 1134 7370278 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000498.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Galt

          Panther Classification:

          Molecular Function -

          nucleotidyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          galactose metabolic process
          UDP-glucose catabolic process
          galactose binding
          glucose binding
          UDP-glucose:hexose-1-phosphate uridylyltransferase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1053772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920272 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1053772 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1053772 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1053772 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1053772 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868397 - -
          Affymetrix Rat Gene 1.0 ST Array 10868387 - -
          Affymetrix Rat Gene 1.0 ST Array 10868395 - -
          Affymetrix Rat Gene 1.0 ST Array 10868390 - -
          Affymetrix Rat Gene 1.0 ST Array 10868394 - -
          Affymetrix Rat Gene 1.0 ST Array 10868389 - -
          Affymetrix Rat Gene 1.0 ST Array 10868385 - -
          Affymetrix Rat Gene 1.0 ST Array 10868388 - -
          Affymetrix Rat Gene 1.0 ST Array 10868386 - -
          Affymetrix Rat Gene 1.0 ST Array 10868396 - -
          Affymetrix Rat Gene 1.0 ST Array 10868391 - -
          Affymetrix Rat Gene 1.0 ST Array 10868392 - -
          Affymetrix Rat Gene 1.0 ST Array 10868393 - -
          Affymetrix Rat Gene 1.1 ST Array 10868384 - -
          Rat Genome 230 2.0 Array 1371251_at - -
          Rat Genome 230 2.0 Array 1371251_PM_at - -
          Affymetrix HT Rat Focus Array 1371251_PM_at - -
          Affymetrix HT Rat Focus Array 1371251_at - -
          Affymetrix Rat Clariom S Assay PSR0500003035.rn.1 TC0500000498.rn.1 RTA TC
          Rat Expression Set 230 1371251_at - -
          Rat Expression Set 230 1371251_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10868384 - -
          Rat Genome U34 Set L05541_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20870257 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357759 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline