Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01765565_m1
          See other BRK1 GE Assays ›
          Gene Symbol:
          Brk1
          Gene Name:
          BRICK1, SCAR/WAVE actin-nucleating complex subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 145559206 - 145574438 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01765565_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Brk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195476.1 NP_001182405.1 1 - 2 137 75
          GenBank mRNA FM063685.1 - 1 - 2 125 75
          FQ213048.1 - 1 - 2 137 75
          FQ213578.1 - 1 - 2 137 75
          FQ214114.1 - 1 - 2 137 75
          FQ220806.1 - 1 - 2 135 75
          FQ226823.1 - 1 - 2 137 75
          FQ229010.1 - 1 - 2 137 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001578.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          positive regulation of cell proliferation
          positive regulation of lamellipodium assembly
          Rac protein signal transduction
          actin cytoskeleton organization
          cell motility
          protein homotrimerization
          positive regulation of Arp2/3 complex-mediated actin nucleation
          protein complex binding
          Rac GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133343 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P496375 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133343 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P496375 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P133343 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P496375 - -
          Array Name Probe IDs Targets Target Type
          Rat Genome 230 2.0 Array 1388854_PM_at - -
          Rat Genome 230 2.0 Array 1388854_at - -
          Affymetrix HT Rat Focus Array 1388854_PM_at - -
          Affymetrix HT Rat Focus Array 1388854_at - -
          Affymetrix Rat Clariom S Assay PSR0400011239.rn.1 TC0400001578.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011238.rn.1 TC0400001578.rn.1 RTA TC
          Rat Expression Set 230 1388854_at - -
          Rat Expression Set 230 1388854_PM_at - -
          Rat Genome U34 Set rc_AA944169_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20851351 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline