Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767215_m1
          See other GABPA GE Assays ›
          Gene Symbol:
          Gabpa
          Gene Name:
          GA binding protein transcription factor, alpha subunit
          Gene Aliases:
          Nrf-2alpha, Nrf2a
          Chromosome Location:
          Chr.11: 24293933 - 24322822 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01767215_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gabpa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108841.1 NP_001102311.1 1226 74
          XM_008768584.1 XP_008766806.1 6 - 7 835 74
          XM_008768585.1 XP_008766807.1 6 - 7 830 74
          XM_008768586.1 XP_008766808.1 6 - 7 830 74
          XM_008768587.1 XP_008766809.1 5 - 6 640 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000153.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell differentiation
          negative regulation of megakaryocyte differentiation
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription regulatory region DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P739060 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P359190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160753 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55159 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P359190 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P739060 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P359190 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P739060 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P739060 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P359190 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P528302 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P359190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90460 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5092 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750036 - -
          Affymetrix Rat Gene 1.0 ST Array 10750037 - -
          Affymetrix Rat Gene 1.0 ST Array 10750031 - -
          Affymetrix Rat Gene 1.0 ST Array 10750032 - -
          Affymetrix Rat Gene 1.0 ST Array 10750033 - -
          Affymetrix Rat Gene 1.0 ST Array 10750038 - -
          Affymetrix Rat Gene 1.0 ST Array 10750034 - -
          Affymetrix Rat Gene 1.0 ST Array 10750035 - -
          Affymetrix Rat Gene 1.0 ST Array 10750039 - -
          Affymetrix Rat Gene 1.1 ST Array 10750029 - -
          Rat Genome 230 2.0 Array 1383377_PM_at - -
          Rat Genome 230 2.0 Array 1383377_at - -
          Affymetrix HT Rat Focus Array 1383377_PM_at - -
          Affymetrix HT Rat Focus Array 1383377_at - -
          Affymetrix Rat Clariom S Assay PSR1100000546.rn.1 TC1100000153.rn.1 RTA TC
          Rat Expression Set 230 1383377_PM_at - -
          Rat Expression Set 230 1383377_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750029 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22098305 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355153 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
          git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.51.0-2026.05.21-1.0