Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767244_m1
          See other SON GE Assays ›
          Gene Symbol:
          Son
          Gene Name:
          Son DNA binding protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 31806600 - 31837769 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01767244_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Son
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170327.1 NP_001163798.1 2 - 3 295 99
          NM_001170328.1 NP_001163799.1 2 - 3 295 99
          XM_006248064.2 XP_006248126.1 2 - 3 313 99
          XM_006248065.2 XP_006248127.1 2 - 3 313 99
          XM_008768580.1 XP_008766802.1 2 - 3 313 99
          GenBank mRNA BC058506.1 - 2 - 3 314 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000256.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          mRNA processing
          negative regulation of apoptotic process
          regulation of RNA splicing
          regulation of cell cycle
          RNA binding
          poly(A) RNA binding
          RS domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1100001020.rn.1 TC1100000256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001012.rn.1 TC1100000256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001014.rn.1 TC1100000256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001016.rn.1 TC1100000256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001013.rn.1 TC1100000256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001015.rn.1 TC1100000256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001018.rn.1 TC1100000256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001017.rn.1 TC1100000256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001019.rn.1 TC1100000256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001021.rn.1 TC1100000256.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001022.rn.1 TC1100000256.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline