Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767331_m1
          See other ROBO3 GE Assays ›
          Gene Symbol:
          Robo3
          Gene Name:
          roundabout, axon guidance receptor, homolog 3 (Drosophila)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 39892792 - 39910596 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01767331_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Robo3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108135.1 NP_001101605.1 6 - 7 1323 73
          XM_008766057.1 XP_008764279.1 7 - 8 1448 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002263.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          neuron migration
          axon guidance
          axon midline choice point recognition
          chemoattraction of axon
          chemorepulsion of axon
          commissural neuron axon guidance

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P041937 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P608485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851537 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88359 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170348 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041937 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P608485 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P608485 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041937 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P041937 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P608485 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P149977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39297 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10916205 - -
          Affymetrix Rat Gene 1.0 ST Array 10916220 - -
          Affymetrix Rat Gene 1.0 ST Array 10916224 - -
          Affymetrix Rat Gene 1.0 ST Array 10916221 - -
          Affymetrix Rat Gene 1.0 ST Array 10916222 - -
          Affymetrix Rat Gene 1.0 ST Array 10916226 - -
          Affymetrix Rat Gene 1.0 ST Array 10916217 - -
          Affymetrix Rat Gene 1.0 ST Array 10916206 - -
          Affymetrix Rat Gene 1.0 ST Array 10916225 - -
          Affymetrix Rat Gene 1.0 ST Array 10916214 - -
          Affymetrix Rat Gene 1.0 ST Array 10916211 - -
          Affymetrix Rat Gene 1.0 ST Array 10916207 - -
          Affymetrix Rat Gene 1.0 ST Array 10916203 - -
          Affymetrix Rat Gene 1.0 ST Array 10916213 - -
          Affymetrix Rat Gene 1.0 ST Array 10916204 - -
          Affymetrix Rat Gene 1.0 ST Array 10916208 - -
          Affymetrix Rat Gene 1.0 ST Array 10916219 - -
          Affymetrix Rat Gene 1.0 ST Array 10916209 - -
          Affymetrix Rat Gene 1.0 ST Array 10916223 - -
          Affymetrix Rat Gene 1.0 ST Array 10916202 - -
          Affymetrix Rat Gene 1.0 ST Array 10916218 - -
          Affymetrix Rat Gene 1.0 ST Array 10916215 - -
          Affymetrix Rat Gene 1.0 ST Array 10916212 - -
          Affymetrix Rat Gene 1.0 ST Array 10916216 - -
          Affymetrix Rat Gene 1.0 ST Array 10916210 - -
          Affymetrix Rat Gene 1.1 ST Array 10916201 - -
          Rat Genome 230 2.0 Array 1380370_PM_at - -
          Rat Genome 230 2.0 Array 1380370_at - -
          Affymetrix HT Rat Focus Array 1380370_at - -
          Affymetrix HT Rat Focus Array 1380370_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800018225.rn.1 TC0800002263.rn.1 RTA TC
          Rat Expression Set 230 1380370_PM_at - -
          Rat Expression Set 230 1380370_at - -
          Affymetrix Rat Gene 1.0 ST Array 10916201 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353622 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline