Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767850_m1
          See other RANGAP1 GE Assays ›
          Gene Symbol:
          Rangap1
          Gene Name:
          RAN GTPase activating protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 122939263 - 122967178 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01767850_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rangap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001244855.1 NP_001231784.1 13 - 14 1698 72
          XM_006242100.2 XP_006242162.1 13 - 14 3138 72
          XM_006242101.2 XP_006242163.1 13 - 14 1987 72
          GenBank mRNA BC082056.1 - 13 - 14 1606 7132303 72
          BP492086.1 - 3 - 4 249 72
          FQ217036.1 - 13 - 14 1682 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003552.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s)

          signal transduction
          negative regulation of protein export from nucleus

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P166495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14305 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P166495 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P166495 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P166495 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P166495 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P166501 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44761 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43333 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101934 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10905636 - -
          Affymetrix Rat Gene 1.0 ST Array 10905641 - -
          Affymetrix Rat Gene 1.0 ST Array 10905629 - -
          Affymetrix Rat Gene 1.0 ST Array 10905643 - -
          Affymetrix Rat Gene 1.0 ST Array 10905638 - -
          Affymetrix Rat Gene 1.0 ST Array 10905642 - -
          Affymetrix Rat Gene 1.0 ST Array 10905628 - -
          Affymetrix Rat Gene 1.0 ST Array 10905634 - -
          Affymetrix Rat Gene 1.0 ST Array 10905630 - -
          Affymetrix Rat Gene 1.0 ST Array 10905640 - -
          Affymetrix Rat Gene 1.0 ST Array 10905635 - -
          Affymetrix Rat Gene 1.0 ST Array 10905633 - -
          Affymetrix Rat Gene 1.0 ST Array 10905639 - -
          Affymetrix Rat Gene 1.0 ST Array 10905637 - -
          Affymetrix Rat Gene 1.0 ST Array 10905631 - -
          Affymetrix Rat Gene 1.0 ST Array 10905627 - -
          Affymetrix Rat Gene 1.1 ST Array 10905625 - -
          Rat Genome 230 2.0 Array 1397465_PM_at - -
          Rat Genome 230 2.0 Array 1388341_PM_at - -
          Rat Genome 230 2.0 Array 1388341_at - -
          Rat Genome 230 2.0 Array 1397465_at - -
          Affymetrix HT Rat Focus Array 1388341_at - -
          Affymetrix HT Rat Focus Array 1388341_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700027986.rn.1 TC0700003552.rn.1 RTA TC
          Rat Expression Set 230 1388341_PM_at - -
          Rat Expression Set 230 1388341_at - -
          Rat Expression Set 230 1397465_PM_at - -
          Rat Expression Set 230 1397465_at - -
          Affymetrix Rat Gene 1.0 ST Array 10905625 - -
          Rat Genome U34 Set rc_AA946189_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21326125 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375936 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376310 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline