Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01769237_m1
          See other TMEM214 GE Assays ›
          Gene Symbol:
          Tmem214
          Gene Name:
          transmembrane protein 214
          Gene Aliases:
          RGD1311324
          Chromosome Location:
          Chr.6: 26867638 - 26875384 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01769237_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem214
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014195.2 NP_001014217.2 13 - 14 1572 66
          GenBank mRNA BC085114.1 - 10 - 11 1003 7106107 66
          BC129115.1 - 13 - 14 1572 7112909 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003496.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem214

          Gene Ontology Categories:

          Function(s)

          apoptotic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P140193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559673 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140193 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P140193 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P201726 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10888982 - -
          Affymetrix Rat Gene 1.0 ST Array 10888988 - -
          Affymetrix Rat Gene 1.0 ST Array 10888990 - -
          Affymetrix Rat Gene 1.0 ST Array 10888994 - -
          Affymetrix Rat Gene 1.0 ST Array 10888991 - -
          Affymetrix Rat Gene 1.0 ST Array 10888987 - -
          Affymetrix Rat Gene 1.0 ST Array 10888985 - -
          Affymetrix Rat Gene 1.0 ST Array 10888984 - -
          Affymetrix Rat Gene 1.0 ST Array 10888995 - -
          Affymetrix Rat Gene 1.0 ST Array 10888993 - -
          Affymetrix Rat Gene 1.0 ST Array 10888981 - -
          Affymetrix Rat Gene 1.0 ST Array 10888983 - -
          Affymetrix Rat Gene 1.0 ST Array 10888989 - -
          Affymetrix Rat Gene 1.0 ST Array 10888986 - -
          Affymetrix Rat Gene 1.0 ST Array 10888992 - -
          Affymetrix Rat Gene 1.0 ST Array 10888997 - -
          Affymetrix Rat Gene 1.0 ST Array 10888996 - -
          Affymetrix Rat Gene 1.1 ST Array 10888980 - -
          Rat Genome 230 2.0 Array 1382596_PM_a_at - -
          Rat Genome 230 2.0 Array 1382596_a_at - -
          Affymetrix HT Rat Focus Array 1382596_PM_a_at - -
          Affymetrix HT Rat Focus Array 1382596_a_at - -
          Affymetrix Rat Clariom S Assay PSR0600024678.rn.1 TC0600003496.rn.1 RTA TC
          Rat Expression Set 230 1382596_PM_a_at - -
          Rat Expression Set 230 1382596_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10888980 - -
          Rat Genome U34 Set rc_AA866454_at - -
          Rat Genome U34 Set rc_AA866454_g_at - -
          Rat Genome U34 Set rc_AI230388_at - -
          Rat Genome U34 Set rc_AI172523_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22360685 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357101 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline