Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01771476_m1
          See other KANSL3 GE Assays ›
          Gene Symbol:
          Kansl3
          Gene Name:
          KAT8 regulatory NSL complex subunit 3
          Gene Aliases:
          RGD1309220
          Chromosome Location:
          Chr.9: 42899432 - 42943213 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01771476_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kansl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034835.1 NP_001030007.1 15 - 16 2215 81
          XM_006244808.2 XP_006244870.1 17 - 18 2354 81
          XM_006244809.2 XP_006244871.1 17 - 18 2407 81
          XM_006244810.2 XP_006244872.1 17 - 18 2585 81
          XM_006244811.1 XP_006244873.1 17 - 18 2321 81
          XM_006244812.1 XP_006244874.1 16 - 17 2068 81
          XM_006244813.1 XP_006244875.1 16 - 17 1973 81
          XM_008767027.1 XP_008765249.1 17 - 18 2349 81
          XM_008767028.1 XP_008765250.1 15 - 16 2196 81
          GenBank mRNA BC105865.1 - 15 - 16 2215 7461395 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001815.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kansl3

          Gene Ontology Categories:

          Function(s) Process(es)

          histone H4-K5 acetylation
          histone H4-K8 acetylation
          histone H4-K16 acetylation
          histone acetyltransferase activity (H4-K5 specific)
          histone acetyltransferase activity (H4-K8 specific)
          histone acetyltransferase activity (H4-K16 specific)

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P975235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88201 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121197 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6558 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66436 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P975235 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P975235 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P975235 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P466827 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P975235 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5664 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4817 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21653 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10927341 - -
          Affymetrix Rat Gene 1.0 ST Array 10927351 - -
          Affymetrix Rat Gene 1.0 ST Array 10927348 - -
          Affymetrix Rat Gene 1.0 ST Array 10927350 - -
          Affymetrix Rat Gene 1.0 ST Array 10927335 - -
          Affymetrix Rat Gene 1.0 ST Array 10927349 - -
          Affymetrix Rat Gene 1.0 ST Array 10927344 - -
          Affymetrix Rat Gene 1.0 ST Array 10927339 - -
          Affymetrix Rat Gene 1.0 ST Array 10927336 - -
          Affymetrix Rat Gene 1.0 ST Array 10927337 - -
          Affymetrix Rat Gene 1.0 ST Array 10927342 - -
          Affymetrix Rat Gene 1.0 ST Array 10927345 - -
          Affymetrix Rat Gene 1.0 ST Array 10927334 - -
          Affymetrix Rat Gene 1.0 ST Array 10927347 - -
          Affymetrix Rat Gene 1.0 ST Array 10927346 - -
          Affymetrix Rat Gene 1.0 ST Array 10927343 - -
          Affymetrix Rat Gene 1.0 ST Array 10927338 - -
          Affymetrix Rat Gene 1.0 ST Array 10927333 - -
          Affymetrix Rat Gene 1.0 ST Array 10927340 - -
          Affymetrix Rat Gene 1.1 ST Array 10927332 - -
          Rat Genome 230 2.0 Array 1388637_at - -
          Rat Genome 230 2.0 Array 1388637_PM_at - -
          Affymetrix HT Rat Focus Array 1388637_PM_at - -
          Affymetrix HT Rat Focus Array 1388637_at - -
          Affymetrix Rat Clariom S Assay PSR0900014797.rn.1 TC0900001815.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900014799.rn.1 TC0900001815.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900014798.rn.1 TC0900001815.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900014800.rn.1 TC0900001815.rn.1 RTA TC
          Rat Expression Set 230 1388637_PM_at - -
          Rat Expression Set 230 1388637_at - -
          Affymetrix Rat Gene 1.0 ST Array 10927332 - -
          Rat Genome U34 Set rc_AI144895_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21282621 - -
          ABI Rat Genome Survey Microarray 22340433 - -
          ABI Rat Genome Survey Microarray 22414870 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357357 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline