Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01775924_s1
          See other TP53RK GE Assays ›
          Gene Symbol:
          Tp53rk
          Gene Name:
          TP53 regulating kinase
          Gene Aliases:
          Trp53rk
          Chromosome Location:
          Chr.3: 163552166 - 163555925 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01775924_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tp53rk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108606.1 NP_001102076.1 2062 72

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 Protease 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          tRNA threonylcarbamoyladenosine metabolic process
          p53 binding
          protein serine/threonine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P035228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662546 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035228 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P035228 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P389400 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842343 - -
          Affymetrix Rat Gene 1.1 ST Array 10842341 - -
          Rat Genome 230 2.0 Array 1392417_PM_s_at - -
          Rat Genome 230 2.0 Array 1381822_PM_x_at - -
          Rat Genome 230 2.0 Array 1379488_at - -
          Rat Genome 230 2.0 Array 1379488_PM_at - -
          Rat Genome 230 2.0 Array 1392417_s_at - -
          Rat Genome 230 2.0 Array 1381822_x_at - -
          Affymetrix HT Rat Focus Array 1379488_PM_at - -
          Affymetrix HT Rat Focus Array 1379488_at - -
          Affymetrix Rat Clariom S Assay PSR0300017509.rn.1 TC0300002350.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017510.rn.1 TC0300002350.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017508.rn.1 TC0300002350.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017504.rn.1 TC0300002350.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017503.rn.1 TC0300002350.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017507.rn.1 TC0300002350.rn.1 RTA TC
          Rat Expression Set 230 1381822_x_at - -
          Rat Expression Set 230 1379488_PM_at - -
          Rat Expression Set 230 1379488_at - -
          Rat Expression Set 230 1392417_PM_s_at - -
          Rat Expression Set 230 1392417_s_at - -
          Rat Expression Set 230 1381822_PM_x_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842341 - -
          Rat Genome U34 Set rc_AI071023_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20727773 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline