Search
Search

Olr1l
LOC103689958| Gene Symbol: Olr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001000538.1 | NP_001000538.1 | 1 | 918 | 61 | |
| Gene Symbol: LOC103689958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008758874.1 | XP_008757096.1 | 1 | 918 | 61 | |
Set Membership: |
|