Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02134090_s1
          See other CXCR3 GE Assays ›
          Gene Symbol:
          Cxcr3
          Gene Name:
          chemokine (C-X-C motif) receptor 3
          Gene Aliases:
          Gpr9
          Chromosome Location:
          Chr.X: 71614346 - 71616997 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02134090_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxcr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053415.1 NP_445867.1 2 1610 124
          GenBank mRNA AF223642.1 - 2 1610 124
          BC091136.1 - 1 1465 7376065 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxcr3

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          negative regulation of endothelial cell proliferation
          regulation of leukocyte migration
          chemotaxis
          inflammatory response
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          positive regulation of cell proliferation
          T cell chemotaxis
          negative regulation of angiogenesis
          sensory perception of pain
          calcium-mediated signaling
          positive regulation of cAMP metabolic process
          positive regulation of cAMP-mediated signaling
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of chemotaxis
          positive regulation of release of sequestered calcium ion into cytosol
          chemokine-mediated signaling pathway
          chemokine (C-C motif) ligand 11 production
          negative regulation of execution phase of apoptosis
          positive regulation of execution phase of apoptosis
          receptor activity
          chemokine receptor activity
          C-X-C chemokine receptor activity
          chemokine binding
          C-X-C chemokine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P404843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800975 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P404843 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P404843 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P404843 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P404843 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702446320 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86948 - -
          Agilent Rat cDNA Microarray (G4105A) 702446320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938706 - -
          Affymetrix Rat Gene 1.0 ST Array 10938704 - -
          Affymetrix Rat Gene 1.1 ST Array 10938703 - -
          Rat Genome 230 2.0 Array 1368192_at - -
          Rat Genome 230 2.0 Array 1368192_PM_at - -
          Affymetrix HT Rat Focus Array 1368192_PM_at - -
          Affymetrix HT Rat Focus Array 1368192_at - -
          Rat Expression Set 230 1368192_PM_at - -
          Rat Expression Set 230 1368192_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938703 - -
          Rat Genome U34 Set rc_AI234161_at - -
          Rat Genome U34 Set rc_AI072601_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22099747 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355853 - -
          RatRef-12 Expression BeadChip ILMN_68427 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline