Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02350111_gH
          See other RT1-CE10 GE Assays ›
          Gene Symbol:
          RT1-CE10
          Gene Name:
          RT1 class I, locus CE10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 154072 - 157679 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02350111_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RT1-CE10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008833.1 NP_001008833.1 4 - 5 895 114
          XM_006255847.2 XP_006255909.1 4 - 5 900 114
          GenBank mRNA BC083874.1 - 3 - 4 1079 114
          BC098952.1 - 4 - 5 898 7366301 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000925.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RT1-CE10

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          antigen processing and presentation of peptide antigen via MHC class I
          immune response
          receptor binding
          peptide antigen binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P389019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P389019 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P389019 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P389019 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P389019 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828007 - -
          Affymetrix Rat Gene 1.0 ST Array 10828006 - -
          Affymetrix Rat Gene 1.0 ST Array 10828004 - -
          Affymetrix Rat Gene 1.0 ST Array 10828005 - -
          Affymetrix Rat Gene 1.0 ST Array 10828003 - -
          Affymetrix Rat Gene 1.0 ST Array 10828009 - -
          Affymetrix Rat Gene 1.0 ST Array 10828008 - -
          Affymetrix Rat Gene 1.0 ST Array 10828002 - -
          Affymetrix Rat Gene 1.1 ST Array 10828001 - -
          Rat Genome 230 2.0 Array 1388203_PM_x_at - -
          Rat Genome 230 2.0 Array 1388203_x_at - -
          Affymetrix Rat Clariom S Assay PSR2000007571.rn.1 TC2000000925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000007568.rn.1 TC2000000925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000007572.rn.1 TC2000000925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000007570.rn.1 TC2000000925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000007566.rn.1 TC2000000925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000007567.rn.1 TC2000000925.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000007565.rn.1 TC2000000925.rn.1 RTA TC
          Rat Expression Set 230 1388203_x_at - -
          Rat Expression Set 230 1388203_PM_x_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828001 - -
          Rat Genome U34 Set rc_AI103500_f_at - -
          Rat Genome U34 Set rc_AI103500_i_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline