Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02395596_g1
          See other LOC363337 GE Assays ›
          Gene Symbol:
          LOC363337
          Gene Name:
          similar to RIKEN cDNA 1700081O22
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 27858878 - 27862883 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02395596_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC363337
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014221.1 NP_001014243.1 4 - 5 1343 124
          XM_006255363.2 XP_006255425.1 5 - 6 766 124
          GenBank mRNA BC082068.1 - 4 - 5 1343 7132781 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC363337

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P229677 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P124898 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P069789 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P046885 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P100208 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P012477 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P035237 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100208 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069789 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046885 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P229677 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012477 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P124898 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035237 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P124898 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035237 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P229677 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069789 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046885 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012477 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100208 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P069789 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P124898 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P229677 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P035237 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P012477 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P046885 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P100208 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P140225 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P171155 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P229677 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P255728 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P124121 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P135786 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P149896 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P114352 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58046 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66280 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103525 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32519 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10931086 - -
          Affymetrix Rat Gene 1.0 ST Array 10931085 - -
          Affymetrix Rat Gene 1.0 ST Array 10775257 - -
          Affymetrix Rat Gene 1.0 ST Array 10775248 - -
          Affymetrix Rat Gene 1.0 ST Array 10775261 - -
          Affymetrix Rat Gene 1.0 ST Array 10931087 - -
          Affymetrix Rat Gene 1.0 ST Array 10931084 - -
          Affymetrix Rat Gene 1.0 ST Array 10775249 - -
          Affymetrix Rat Gene 1.0 ST Array 10806199 - -
          Affymetrix Rat Gene 1.1 ST Array 10775243 - -
          Affymetrix Rat Gene 1.1 ST Array 10806198 - -
          Affymetrix Rat Gene 1.1 ST Array 10931083 - -
          Affymetrix Rat Gene 1.1 ST Array 10775260 - -
          Affymetrix Rat Clariom S Assay PSR1900010121.rn.1 TC1900001245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900010119.rn.1 TC1900001245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900010120.rn.1 TC1900001245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900010122.rn.1 TC1900001245.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10931083 - -
          Rat Genome U34 Set rc_AI228425_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20999277 - -
          ABI Rat Genome Survey Microarray 21242921 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352819 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline