Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02532935_s1
          See other RGD1308147 GE Assays ›
          Gene Symbol:
          RGD1308147
          Gene Name:
          similar to expressed sequence AW209491
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 53080634 - 53087274 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02532935_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1308147
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014010.1 NP_001014032.1 2 1573 67
          GenBank mRNA BC088171.1 - 2 1573 7322765 67
          BC090337.1 - 1 1271 7302860 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RGD1308147

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P538989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21647 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P538989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P538989 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P538989 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21901 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P538989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64760 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798725 - -
          Affymetrix Rat Gene 1.0 ST Array 10798727 - -
          Affymetrix Rat Gene 1.0 ST Array 10798726 - -
          Affymetrix Rat Gene 1.1 ST Array 10798724 - -
          Rat Genome 230 2.0 Array 1373547_PM_at - -
          Rat Genome 230 2.0 Array 1373547_at - -
          Affymetrix HT Rat Focus Array 1373547_PM_at - -
          Affymetrix HT Rat Focus Array 1373547_at - -
          Affymetrix Rat Clariom S Assay PSR1700012922.rn.1 TC1700001917.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700012919.rn.1 TC1700001917.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700012920.rn.1 TC1700001917.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700012923.rn.1 TC1700001917.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700012924.rn.1 TC1700001917.rn.1 RTA TC
          Rat Expression Set 230 1373547_at - -
          Rat Expression Set 230 1373547_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10798724 - -
          Rat Genome U34 Set rc_AI233244_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21389654 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline