Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02769617_s1
          See other FBXO46 GE Assays ›
          Gene Symbol:
          Fbxo46
          Gene Name:
          F-box protein 46
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 80028875 - 80045666 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02769617_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxo46
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025642.1 NP_001020813.1 2 1845 91
          XM_006228399.2 XP_006228461.1 3 2032 91
          XM_006228400.2 XP_006228462.1 3 2644 91
          XM_006228401.2 XP_006228463.1 2 2527 91
          XM_008758910.1 XP_008757132.1 2 2176 91
          GenBank mRNA BC098945.1 - 2 1845 7458607 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxo46

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P458137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164878 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100525 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P458137 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P458137 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P458137 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421757 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 659 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56445 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704753 - -
          Affymetrix Rat Gene 1.0 ST Array 10704754 - -
          Affymetrix Rat Gene 1.1 ST Array 10704752 - -
          Rat Genome 230 2.0 Array 1378266_PM_at - -
          Rat Genome 230 2.0 Array 1378266_at - -
          Affymetrix HT Rat Focus Array 1378266_at - -
          Affymetrix HT Rat Focus Array 1378266_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100006912.rn.1 TC0100000988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006913.rn.1 TC0100000988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006915.rn.1 TC0100000988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006911.rn.1 TC0100000988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006903.rn.1 TC0100000988.rn.1 RTA TC
          Rat Expression Set 230 1378266_at - -
          Rat Expression Set 230 1378266_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704752 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21147767 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366206 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline