Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02769715_g1
          See other LOC303448 GE Assays ›
          Gene Symbol:
          LOC303448
          Gene Name:
          similar to glyceraldehyde-3-phosphate dehydrogenase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 78687653 - 78696097 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02769715_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC303448
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037190.1 NP_001032267.1 2 164 75
          GenBank mRNA BC100150.1 - 2 164 7455257 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC303448

          Panther Classification:

          Molecular Function -

          dehydrogenase

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P027852 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P057453 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P027858 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P045568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912471 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177439 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027858 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045568 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057453 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045568 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027858 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057453 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P045568 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P057453 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P027858 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P027852 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23058 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10746245 - -
          Affymetrix Rat Gene 1.0 ST Array 10746244 - -
          Affymetrix Rat Gene 1.1 ST Array 10754880 - -
          Affymetrix Rat Gene 1.1 ST Array 10746243 - -
          Affymetrix Rat Gene 1.1 ST Array 10840272 - -
          Affymetrix Rat Gene 1.1 ST Array 10802513 - -
          Affymetrix Rat Gene 1.1 ST Array 10909368 - -
          Affymetrix Rat Clariom S Assay PSR1000031741.rn.1 TC1000003560.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031742.rn.1 TC1000003560.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031743.rn.1 TC1000003560.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031740.rn.1 TC1000003560.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031739.rn.1 TC1000003560.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031744.rn.1 TC1000003560.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10746243 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363990 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline